N-O ELISA Kits
Alfa Aesar™ Mouse Osteocalcin Enzyme Immunoassay Kit
For measuring levels of osteocalcin in mouse serum and plasma samples
R&D Systems™ Human Nectin-4 Quantikine ELISA Kit
For the quantitative determination of human Nectin-4 concentrations in cell culture supernates, cell lysates, serum, plasma, and urine
R&D Systems™ Human Neuropilin-1 Quantikine ELISA Kit
a 4.5 hour solid-phase ELISA designed to measure human Neuropilin-1 in cell culture supernates, cell lysates, serum, plasma, urine, and human milk.
Novus Biologicals™ Human OX40/TNFRSF4 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
R&D Systems™ Human Neuropilin-1 DuoSet, 5 Plate
Sufficient for at least five 96-well plates
Alfa Aesar™ Osteocalcin, human, mid-tact, Enzyme Immunoassay Kit
Compatible with cultured cell extracts or scaffold cells
Alfa Aesar™ Osteocalcin, human, intact, Enzyme Immunoassay Kit
Compatible with cultured cell extracts or scaffold cells
eBioscience™ Human Osteopontin Platinum ELISA Kit Non-distribution product as customer accommodation.
For the quantitative detection of human Osteopontin.
R&D Systems™ Human Osteocalcin DuoSet ELISA
R&D Systems™ Human Osteocalcin DuoSet ELISA contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant human Osteocalcin.
eBioscience™ Human OSM-R ELISA Ready-SET-Go!™ Kit Non-distribution product as customer accommodation.
Specifically engineered for accurate and precise measurement of human OSM-R. Oncostatin M is a pleiotropic cytokine that belongs to the IL-6 group of cytokines.
Invitrogen™ Human Neprilysin (MME) ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay human Neprilysin (MME) levels in cell culture supernatants, plasma, serum samples.
Novus Biologicals™ Human OX40 Ligand/TNFSF4 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
R&D Systems™ Human Neutrophil Elastase/ELA2 DuoSet ELISA
R&D Systems™ Human Neutrophil Elastase/ELA2 DuoSet ELISA contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant human Neutrophil Elastase (ELA2).
R&D Systems™ Human OX40/TNFRSF4 DuoSet ELISA
R&D Systems™ Human OX40/TNFRSF4 DuoSet ELISA contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant human OX40.
Invitrogen™ Human NGFR ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay human NGFR levels in cell culture supernatants, plasma, serum samples.
  spinner