NGAL ELISA Kits
R&D Systems™ Human Lipocalin-2/NGAL Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure human Lipocalin-2 in cell culture supernates, serum, plasma, urine, and saliva.
R&D Systems™ Human Lipocalin-2/NGAL DuoSet ELISA
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
  spinner