P-R ELISA Kits
R&D Systems™ Human Periostin/OSF-2 DuoSet ELISA Kit Available on GSA/VA Contract for Federal Government customers only.
Sufficient for at least 15 96-well plates
Invitrogen™ eBioscience™ p53 (Total/Phospho) InstantOne™ ELISA Kit Non-distribution product as customer accommodation.
The InstantOne™ ELISA is specifically engineered for accurate measurement of total and phosphorylated human p53 in cell lysates. The InstantOne™ ELISA kit allows for analysis of samples in approximately one hour.
Novus Biologicals™ Rat BMP-4 ELISA Kit (Chemiluminescence)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
R&D Systems™ Total BDNF Quantikine ELISA Kit Available on GSA/VA Contract for Federal Government customers only.
For the quantitative determination of free- and Trk-bound Brain-Derived Neurotrophic Factor (Total BDNF). The Quantikine Total BDNF Immunoassay is a 3.5 hour solid phase ELISA designed to measure BDNF in cell culture supernates, tissue lysates, serum, plasma, urine, and human milk.
R&D Systems™ Human PSMA/FOLH1 DuoSet ELISA
R&D Systems™ Human PSMA/FOLH1 DuoSet ELISA contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant human Prostate-Specific Membrane Antigen (PSMA).
R&D Systems™ Human Amyloid beta (aa1-40) Quantikine ELISA Kit
For the quantitative determination of human Amyloid beta (amino acids 1-40) concentrations in cell culture supernates, tissue lysates, and cerebrospinal fluid. Human Amyloid beta (aa1-40) Quantikine ELISA Kit employs the quantitative sandwich enzyme immunoassay technique.
eBioscience™ Human PAI-1 Platinum ELISA Kit Non-distribution product as customer accommodation.
For the quantitative detection of human PAI-1.
Invitrogen™ PKA (Protein Kinase A) Colorimetric Activity Kit Non-distribution product as customer accommodation.
Assay PKA (Protein Kinase A) Activity in Cell Lysates, Tissue Extracts and Buffer Samples.
Invitrogen™ Mouse PLGF-2 (PGF) ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay mouse PLGF-2 (PGF) levels in cell culture supernatants, plasma, serum samples
Invitrogen™ Human PSA total (KLK3) ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay human PSA total (KLK3) levels in cell culture supernatants, plasma, serum samples
R&D Systems™ Quantikine Immunoassay Control Group 237 Available on GSA/VA Contract for Federal Government customers only.
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Novus Biologicals™ Human/Mouse/Rat RelA/NFkB p65 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Novus Biologicals™ Porcine Rotavirus Antigen ELISA Kit (Colorimetric)
Assay has high sensitivity and specificity for detection of pig Rotavirus antigen
Novus Biologicals™ Rat AIF-1/Iba1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
R&D Systems™ Human P-Selectin/CD62P Quantikine ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
R&D Systems™ Human Amyloid beta (aa1-42) Quantikine ELISA Kit
For the quantitative determination of human Amyloid beta (amino acids 1-40) concentrations in cell culture supernates, tissue lysates, and cerebrospinal fluid. Human Amyloid beta (aa1-42) Quantikine ELISA Kit employs the quantitative sandwich enzyme immunoassay technique.
Invitrogen™ Mouse Renin 1 (REN1) ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay mouse Renin 1 (REN1) levels in cell culture supernatants, plasma, serum samples.
Invitrogen™ Human Procalcitonin ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay human Procalcitonin levels in cell culture supernatants, plasma, serum samples.
Invitrogen™ Retinol Binding Protein Urinary Competitive ELISA Kit Non-distribution product as customer accommodation.
Assay Retinol Binding Protein levels in Human, Rat, Dog and Rhesus Monkey Urine samples.
Invitrogen™ Human PAPP-A ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay human PAPP-A levels in cell culture supernatants, plasma, serum samples.
R&D Systems™ Human Hepcidin Quantikine ELISA Kit Available on GSA/VA Contract for Federal Government customers only.
For the quantitative determination of human Hepcidin concentrations in cell culture supernates, serum, plasma, and urine. Human Hepcidin Immunoassay employs the quantitative sandwich enzyme immunoassay technique.
Invitrogen™ Canine PDGF-BB ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay canine PDGF-BB levels in cell culture supernatants, plasma, serum samples
Invitrogen™ Rat RAGE (AGER) ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay rat RAGE (AGER) levels in cell culture supernatants, plasma, serum samples
Invitrogen™ GLP-1 Multispecies ELISA Kit Non-distribution product as customer accommodation.
The Human Glucagon Like Peptide-1 (Hu GLP-1) ELISA quantitates Hu GLP-1 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu GLP-1.
R&D Systems™ Rat IL-1 beta/IL-1F2 DuoSet ELISA
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
R&D Systems™ Mouse/Rat PDGF-BB DuoSet™
For the development of sandwich ELISAs to measure natural and recombinant mouse and rat Platelet-derived Growth Factor BB (PDGF-BB)
R&D Systems™ Mouse IL-18 BPd DuoSet ELISA Kit
This Mouse IL-18 BPd DuoSet ELISA Development kit contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant mouse Interleukin 18 Binding Protein Isoform d (IL-18 BPd)
Novus Biologicals™ Rat Agrin ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Novus Biologicals™ Rat LBP ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Invitrogen™ Human Periostin ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay human Periostin levels in cell culture supernatants, plasma, serum samples.
  spinner