Substrates for ELISA Detection
Thermo Scientific™ Pierce™ TMB Substrates Available on GSA/VA Contract for Federal Government customers only.
Horseradish peroxidase (HRP) substrates for ELISA that change from blue to yellow upon addition of a sulfuric or phosphoric acid stop solution.
R&D Systems™ Substrate Reagent Pack Available on GSA/VA Contract for Federal Government customers only.
8 vials of Color Reagent A (stabilized hydrogen peroxide) and 8 vials of Color Reagent B (stabilized tetramethylbenzidine)
Thermo Scientific™ SuperSignal™ ELISA Pico Substrate Available on GSA/VA Contract for Federal Government customers only.
HRP substrate optimized to generate an intense light signal and provide exception performance in luminometer-based assays.
Thermo Scientific™ SuperSignal™ ELISA Femto Maximum Sensitivity Substrate Available on GSA/VA Contract for Federal Government customers only.
HRP substrate formulated for superior protein detection and low-end linearity in chemiluminescent ELISA applications.
BD TMB Substrate Reagent Set
For use in BD OptEIA™ ELISA sets. Substrate Reagent A, which is clear and colorless, contains hydrogen peroxide. Substrate Reagent B is colorless to a very light amber solution containing 3,3′, 5,5′ tetramethylbenzidine (TMB) in an organic solvent.
eBioscience™ Super AquaBlue ELISA Substrate Non-distribution product as customer accommodation.
Enhanced ABTS substrate solution for ELISA which provides high signal and sensitivity, low background. Safe, stable and convenient to use. Oxidized by HRP-labeled probes, resulting in soluble, blue-green colored end product, whose absorption can be measured by spectrophotometers at 405nm.
Thermo Scientific™ Pierce™ PNPP Substrates
Powders, tablets, solutions and complete kits of p-nitrophenyl phosphate, a colorimetric, soluble alkaline phosphatase substrate for use in ELISA applications.
R&D Systems™ Glo Substrate Reagent Pack Available on GSA/VA Contract for Federal Government customers only.
for chemiluminescent immunoassay development
Invitrogen™ eBioscience™ TMB ELISA Substrate Solution, 1x Non-distribution product as customer accommodation.
Recommended for ELISA Ready-SET-Go!™ kits. eBioscience™ TMB ELISA Substrate Solution, 1x is a sensitive chromogenic substrate supplied as single ready-to-use solution.
Thermo Scientific™ QuantaRed™ Enhanced Chemifluorescent HRP Substrate
Enable sensitivity comparable to enhanced chemiluminescence (ECL) using the most sensitive fluorescent ELISA substrate for peroxidase (HRP).
Thermo Scientific™ Pierce™ PNPP Substrates
An alkaline phosphatase substrate for ELISA applications
Thermo Scientific™ QuantaBlu™ Fluorogenic Peroxidase Substrate Available on GSA/VA Contract for Federal Government customers only.
Conduct stopped, non-stopped and kinetic assays using this soluble fluorogenic (chemifluorescent) peroxidase substrate for ELISA.
Thermo Scientific™ Pierce™ Substrate Buffers for Soluble Substrates
Optimized, reliable and convenient stock solutions of peroxide for HRP-based assays and diethanolamine for alkaline phosphatase (alk-phos) detection.
Thermo Scientific™ Pierce™ ABTS Substrates
Ready-to-use, water-soluble peroxidase substrate tablets that yield a measurable green end-product for use in ELISA methods.
Invitrogen™ CDP-Star™ Substrate (0.25mM) with Nitro-Block-II™ Enhancer Non-distribution product as customer accommodation.
Ready-to-use chemiluminescent alkaline phosphatase substrate, for protein or nucleic acid blotting on nitrocellulose membranes or solution-based assays.
Thermo Scientific™ Pierce™ OPD Substrates
Water-soluble horseradish peroxidase (HRP) substrate that produces a yellow-orange product detectable at 492nm by ELISA plate readers.
Thermo Scientific™ Pierce™ ONPG Substrate
Preferred colorimetric beta-galactosidase (b-Gal) substrate for ELISA applications.
Invitrogen™ ELISA-Light™ Immunoassay System with CDP-Star™ and Emerald-II™ Substrate/Enhancer Solution Non-distribution product as customer accommodation.
Ultra-sensitive detection of antigens or antibodies in multiple immunoassay formats employing an alkaline phosphatase enzyme label.
Invitrogen™ CSPD™ Substrate (0.4mM) with Sapphire-II™ Enhancer, Size B Non-distribution product as customer accommodation.
Ready-to-use chemiluminescent alkaline phosphatase substrate, for protein or nucleic acid blotting on nitrocellulose membranes or solution-based assays.
Invitrogen™ eBioscience™ ELISA Wash Buffer Non-distribution product as customer accommodation.
Suitable for washing ELISA plate microwells. eBioscience™ ELISA Wash Buffer is supplied in 10 packets of concentrated powder, yielding a total of 10 liters of 1x Wash Buffer.
Invitrogen™ CDP-Star™ Substrate (0.4mM) with Sapphire-II™ Enhancer, Size A Non-distribution product as customer accommodation.
Ready-to-use chemiluminescent alkaline phosphatase substrate, for protein or nucleic acid blotting on nitrocellulose membranes or solution-based assays.
Invitrogen™ CDP-Star™ Substrate (0.4mM) with Emerald-II™ Enhancer, Size A Non-distribution product as customer accommodation.
Ready-to-use chemiluminescent alkaline phosphatase substrate, for protein or nucleic acid blotting on nitrocellulose membranes or solution-based assays.
Invitrogen™ ELISA-Light™ Immunoassay System with CSPD™ and Sapphire-II™ Substrate/Enhancer Solution Non-distribution product as customer accommodation.
Ultra-sensitive detection of antigens or antibodies in multiple immunoassay formats employing an alkaline phosphatase enzyme label.
Invitrogen™ Galacto-Light™ Reaction Buffer Diluent with Galacton-Plus™ Substrate Non-distribution product as customer accommodation.
Established system with extended light emission and high sensitivity for reporter gene assays. Thermo Scientific Galacto-Light Reaction Buffer Diluent and the Galacton-Plus chemiluminescent substrate solution are used with the Galacto-Light Plus Reporter
Invitrogen™ CSPD™ Substrate (0.25mM) with Nitro-Block™ Enhancer Non-distribution product as customer accommodation.
Ready-to-use chemiluminescent alkaline phosphatase substrate, for protein or nucleic acid blotting on nitrocellulose membranes or solution-based assays.
Novus Biologicals™ NovaLume Atto Chemiluminescent Substrate (AP)
NovaLume Atto Chemiluminescent Substrate (AP) is a ready-to-use stabilized 1, 2-dioxetane substrate.
Invitrogen™ CSPD™ Substrate (0.25mM) Non-distribution product as customer accommodation.
Ready-to-use chemiluminescent alkaline phosphatase substrate for protein or nucleic acid blotting on nitrocellulose membranes or solution-based assays. Thermo Scientific CSPD™ Substrate (0.25mM) can also be used in solution-based assays such as immu
Invitrogen™ CSPD™ Substrate (0.4mM) with Emerald-II™ Enhancer, Size A Non-distribution product as customer accommodation.
Ready-to-use chemiluminescent alkaline phosphatase substrate, for protein or nucleic acid blotting on nitrocellulose membranes or solution-based assays.
  spinner