Immunological Sera
Corning™ Human AB Serum Available on GSA/VA Contract for Federal Government customers only.
Human AB serum is used in tissue engineering and cell therapy applications involving human cell types.
Gibco™ Rabbit Serum Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Standard (sterile-filtered)
MP Biomedicals™ Sheep Serum
Sterile, filtered, Lyophilized Serum
MP Biomedicals™ Whole Serum, Chicken, Purified, 2mL
Whole Chicken Serum with buffer salts
MP Biomedicals™ Goat Serum: Normal, Sterile
Suitable replacement for FBS on most cell lines and primary cultures for Biomedical Research
Human Serum, Type AB, Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
For use as growth supplement for cell culture media
MP Biomedicals™ Rat Serum
Sterile, filtered, Lyophilized Serum
Gibco™ Normal Goat Serum
Normal Goat Serum
MP Biomedicals™ Whole Serum, Goat, Purified, 2mL
Whole Goat Serum with buffer salts
MP Biomedicals™ Rabbit Serum
Sterile, Frozen Brown Solution. Combine with BSA to use as a blocking serum in immunocytochemistry and immunohistochemistry.
Southern Biotech Normal Goat Sera
Processed to prevent minimal hemolysis and filtered to 0.2μm
EMD Millipore™ Calbiochem™ Normal Rabbit Serum Available on GSA/VA Contract for Federal Government customers only.
Used in ELISA, Flow Cytometry, Immunoblotting, Immunofluorescence, Immunohistochemistry, Immunoprecipitation for detection of Rabbit Serum. EMD Millipore™ Calbiochem™ Normal Rabbit Serum is normal carrier sera from healthy, non-immunized rabbits.
MP Biomedicals™ Rabbit Serum
Supports cultures of Borrelia burgdorferi, the causative agent of Lyme disease.
MP Biomedicals™ Human Serum, Pooled
Used as a media component to neutralize cell free virus.
MP Biomedicals™ Human Serum, Type AB, Normal Pool
Used as a growth supplement for cell culture media and in other research applications.
Southern Biotech Normal Mouse Sera
Processed to prevent minimal hemolysis and filtered to 0.2μm
Southern Biotech Normal Rabbit Sera
Processed to prevent minimal hemolysis and filtered to 0.2μm
MP Biomedicals™ Mouse Serum
Used for blocking agents and controls in immunoassays.
MP Biomedicals™ Human Serum: Hepatitis C (HCV)-Free
Hepatitis Associated Antigen (HBsAG)–free, HIV–free
MP Biomedicals™ Goat Serum
Suitable alternative to fetal bovine serum in a wide range of cell culture applications such as the isolation and identification of viruses produced in a variety of primary cell lines.
MP Biomedicals™ Goat Serum: Heat Inactivated
Heat Inactivated serum collected from whole blood from healthy individuals.
EMD Millipore™ Calbiochem™ Normal Goat Serum, Sterile Available on GSA/VA Contract for Federal Government customers only.
Suitable for use in radioimmunoassay. EMD Millipore™ Calbiochem™ Normal Goat Serum, Lyophilized Solid is normal carrier sera, produced from pooled lots of sera from healthy, non-immunized goats.
MP Biomedicals™ Human Serum, Type AB
Golden Yellow Liquid. Proven to grow many human cell lines at a faster rate and with a smaller percentage of serum than mixed blood group serum.
EMD Millipore™ Calbiochem™ Normal Goat Serum Available on GSA/VA Contract for Federal Government customers only.
Used as negative control in any application. EMD Millipore™ Calbiochem™ Normal Goat Serum is normal carrier sera, produced from healthy, non-immunized goats.
  spinner