Mouse Sera
Invitrogen™ eBioscience™ Normal Mouse Serum Non-distribution product as customer accommodation.
Can be used to inhibit nonspecific binding of mouse monoclonal antibodies during staining for flow cytometric analysis. eBioscience™ Normal Mouse Serum can also be used in other applications which require normal mouse serum.
MP Biomedicals™ Mouse Serum
Used for blocking agents and controls in immunoassays.
Southern Biotech Normal Mouse Sera
Processed to prevent minimal hemolysis and filtered to 0.2μm
EMD Millipore™ Calbiochem™ Normal Mouse Serum Available on GSA/VA Contract for Federal Government customers only.
Used in ELISA, Flow Cytometry, Immunoblotting, Immunofluorescence, Immunohistochemistry, Immunoprecipitation for the detection of Mouse Serum. EMD Millipore™ Calbiochem™ Normal Mouse Serum is normal carrier sera from healthy, non-immunized mice.
MP Biomedicals™ Whole Serum, Mouse, Purified, 2mL
Whole Mouse Serum with buffer salts
EMD Millipore™ Mouse IgM Available on GSA/VA Contract for Federal Government customers only.
Mouse IgM Polyclonal Antibody control validated for use in ELISA.
EMD Millipore™ Polyclonal Mouse IgG Available on GSA/VA Contract for Federal Government customers only.
Mouse IgG Polyclonal Antibody control validated for use in ELISA.
  spinner