Galactosidase Assay Kits
Thermo Scientific™ Pierce™ Mammalian β-Galactosidase Assay Kit
Lyse cultured mammalian cells and measure beta-galactosidase activity using compatible M-PER Cell Lysis Reagent and colorimetric b-Gal Assay Reagent.
Thermo Scientific™ Pierce™ Yeast β-Galactosidase Assay Kit
Extract protein from yeast two-hybrid reporter systems and measure beta-galactosidase activity using compatible Y-PER Reagent and b-Gal Assay Reagent.
EMD Millipore™ Novagen™ BetaFluor™ β-Galactosidase Assay Kit
Designed for rapid, sensitive measurement of β-galactosidase activity in cell extracts.
EMD Millipore™ Novagen™ BetaRed™ β-Galactosidase Assay Kit
Designed for rapid, sensitive measurement of β-galactosidase activity in cell extracts.
Genlantis™ X-Gal Staining Kit
Rapid and simple assay for determining transfection efficiency with a beta-galactosidase expressing plasmid
  spinner