Immunohistochemistry Kits and Reagents
Thermo Scientific™ Pierce™ LDH Cytotoxicity Assay Kit Available on GSA/VA Contract for Federal Government customers only.
Quantitatively measure lactose dehydrogenase released into the media from damaged cells as a biomarker for cellular cytotoxicity and cytolysis using a colorimetric assay.
Thermo Scientific™ Pierce™ Methanol-free Formaldehyde Ampules Available on GSA/VA Contract for Federal Government customers only.
Vials of high-quality, methanol-free formaldehyde, a general IHC fixative and reversible amine-crosslinking agent for protein and nucleic acid samples.
Invitrogen™ eBioscience™ Fixation/Permeabilization Concentrate Non-distribution product as customer accommodation.
Intended to be used with antibodies to transcription factors and nuclear proteins, such as Foxp3 and Ki-67, as well as cytokines and chemokines.
Thermo Scientific™ Pierce™ Hoechst 3342 Fluorescent Stain Available on GSA/VA Contract for Federal Government customers only.
High-quality Hoechst dye solution for fluorescent staining of DNA and nuclei in fixed- or live-cell imaging techniques.
Thermo Scientific™ Pierce™ DAPI Nuclear Counterstain Available on GSA/VA Contract for Federal Government customers only.
Highly pure diamidino-2-phenylindole (DAPI) dye for fluorescent staining of DNA content and nuclei in fixed-cell imaging techniques.
Thermo Scientific™ Pierce™ Metal Enhanced DAB Substrate Kit Available on GSA/VA Contract for Federal Government customers only.
Special formulation of diaminobenzidine peroxidase substrate that uses cobalt and nickel chloride to yield intense color for IHC staining.
BD ELISPOT Sets Available on GSA/VA Contract for Federal Government customers only.
Sufficient capture antibody, detection antibody, and streptavidin-horseradish peroxidase for 10 × 96 wells
Thermo Scientific™ Pierce™ Protein Transfection Reagent Available on GSA/VA Contract for Federal Government customers only.
Form complexes with proteins, peptides, antibodies and other biologically active molecules for direct intracellular delivery using this cationic lipid mixture.
Thermo Scientific™ Pierce™ ABC Staining Kits Available on GSA/VA Contract for Federal Government customers only.
Detect biotinylated secondary antibodies by IHC using the highly sensitive avidin-biotin complex (ABC) technique.
Novex™ Endogenous Biotin-Blocking Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Endogenous Biotin-Blocking Kit
BD BV480 Mouse Anti-Ki-67
Expressed in the nucleus of cycling cells (G1, S, G2, M cell cycle phases). BD Horizon™ BV480 Mouse Anti-Ki-67 is a large protein having 2 alternatively spliced isoforms, an N-terminal forkhead-associated domain, a C-terminal domain that binds to heterochromatin proteins, and multiple phosphorylation sites, the functions of which are still unclear.
InSure™ FIT™ Immunochemical Test Kit
The InSure FIT test and its unique Blue Brush Method make it easier to screen for colorectal cancer
BD CBA Mouse Inflammation Kit Available on GSA/VA Contract for Federal Government customers only.
Optimized for analysis of specific proteins in tissue culture supernatants, EDTA plasma, and serum samples
Invitrogen™ eBioscience™ IHC Antigen Retrieval Solution, Low pH, 10x Non-distribution product as customer accommodation.
Designed for use during the heat induced epitope retrieval (HIER) step prior to immunohistochemistry on formalin-fixed paraffin-embedded tissue sections. Used in combination with heat has been shown to restore the antigenicity of proteins modified during the formalin fixation of tissue.
Thermo Scientific™ Pierce™ Peroxidase IHC Detection Kit
Bundles the Pierce Metal Enhanced DAB Substrate Kit with all components to stain, counterstain and preserve results from frozen or paraffin tissue sections.
BD Cytometric Bead Array (CBA) Human TNF; Flex Set, Bead D9 Available on GSA/VA Contract for Federal Government customers only.
Bead-based immunoassay capable of measuring human tumor necrosis factor (TNF), formerly known as tumor necrosis factor-a, in serum, plasma, and cell culture supernatant samples
BD BV480 Mouse Anti-Human CD20
CD20 is expressed on pre-B-cells, resting and activated B cells, and follicular dendritic cells, but not plasma cells. BD Horizon™ BV480 Mouse Anti-Human CD20 is observed in low levels on a small subset of normal circulating T lymphocytes. The CD20 molecule is involved in the regulation of B-cell activation.
Meridian Bioscience™ Latex Agglutination System Available on GSA/VA Contract for Federal Government customers only.
Qualitative and semi-quantitative latex test for detection of Cryptococcus neoformans
BD Human IFN-γ ELISA Kit II Available on GSA/VA Contract for Federal Government customers only.
For in vitro quantitative determination of human interferons in serum, plasma, and cell culture samples
BD BV480 Rat Anti-Mouse CD19
Expressed throughout B-lymphocyte development from the pro-B cell through the mature B-cell stages. BD Horizon™ BV480 Rat Anti-Mouse CD19 participates in B-lymphocyte development, B-cell activation, maturation of memory B cells and regulation of tolerance. CD19 has also been detected on peritoneal mast cells, co-localized with CD21/CD35, and it is proposed to play a role in complement-mediated mast-cell activation.
EMD Millipore™ Black Gold II Stain Available on GSA/VA Contract for Federal Government customers only.
Stains specifically for myelin within the central nervous system, provides high resolution, high contrast, a short histochemical processing time, and high reproducibility.
BD BV480 Mouse Anti-Human CD14
CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB. BD Horizon™ BV480 Mouse Anti-Human CD14-specific antibody reacts with interfollicular macrophages, reticular dendritic cells and some Langerhans cells.
BD BV480 Mouse Anti-Human HLA-DR
HLA-DR antigens are encoded by genes within the Human Leukocyte Antigen (HLA) Complex located on chromosome 6. BD Horizon™ BV480 Mouse Anti-Human HLA-DR is also expressed on activated T cells. This molecule plays a major role in mediating cellular interactions during antigen presentation to CD4-positive T cells.
Invitrogen™ Click-iT™ TUNEL Colorimetric IHC Detection Kit Non-distribution product as customer accommodation.
Click-iT TUNEL Colorimetric IHC Detection Kit
Thermo Scientific™ Pierce™ Peroxidase Suppressor
Stable, easy-to-use endogenous peroxidase inhibitor to reduce background signal from cells and tissues in HRP-based IHC staining.
Molecular Probes™ Pluripotent Stem Cell 4-Marker Immunocytochemistry Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Pluripotent Stem Cell 4-Marker Immunocytochemistry Kit
BD BV480 Mouse Anti-Human CD16
Expressed on NK cells as well as macrophages and granulocytes. Reports indicate that CD16 plays a role in signal transduction and NK cell activation. BD Horizon™ BV480 Mouse Anti-Human CD16 clone 3G8 also cross-reacts with a  subset of peripheral blood lymphocytes and monocytes, but not granulocytes, of baboon and both rhesus and cynomologus macaque monkeys.
BD Cytometric Bead Array (CBA) Human IL-6; Flex Set, Bead A7 Available on GSA/VA Contract for Federal Government customers only.
Bead-based immunoassay capable of measuring human interleukin-6 (IL-6) in serum, plasma, and cell culture supernatant samples
  spinner