Mass-Tagged Crosslinkers
Thermo Scientific™ Pierce™ BS3-d0 and BS3-d4 Crosslinker Mass Pair
Crosslink amines to amines using these heavy (deuterated) and light matched pairs of an 11.4A, NHS-ester crosslinker for mass spectrometry analysis of protein interactions.
Thermo Scientific™ Pierce™ BS2G-d0 and BS2G-d4 Crosslinker Mass Pair
Crosslink amines to amines using these heavy (deuterated) and light matched pairs of a 7.7A, NHS-ester crosslinker for mass spectrometry analysis of protein interactions.
  spinner