Miscellaneous Reagents for Molecular Biology
Thermo Scientific™ Pierce™ D-Luciferin Available on GSA/VA Contract for Federal Government customers only.
Firefly luciferase substrate for in vitro and in vivo applications
Thermo Scientific™ IPTG Available on GSA/VA Contract for Federal Government customers only.
Induce the expression of genes under the control of lac operon in cloning and protein expression procedures.
Thermo Scientific™ TransformAid Bacterial Transformation Kit
Kit for quick and convenient preparation of competent E.coli cells for guaranted transformation efficiencies of more than 107 transformants per µg of plasmid DNA.
Thermo Scientific™ X-Gal Solution, ready-to-use Available on GSA/VA Contract for Federal Government customers only.
Effectively screen colonies with X-Gal, an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal, forming an intense blue precipitate.
Thermo Scientific™ DTT Available on GSA/VA Contract for Federal Government customers only.
Stabilize enzymes and other proteins which possess free sulfhydryl groups with DTT, shown to restore activity lost by oxidation of these groups in vitro.
EMD Millipore™ Novagen™ Pellet Paint™ NF Co-Precipitant Available on GSA/VA Contract for Federal Government customers only.
Useful substitute for the original Pellet Paint Co-Precipitant in applications where fluorescent detection is used
EMD Millipore™ Calbiochem™ Tetrahydrouridine Available on GSA/VA Contract for Federal Government customers only.
Potent competitive inhibitor of cytidine deaminase
Invitrogen™ IPTG Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Inducer of β-galactosidase activity in bacteria and is suitable for use with X-gal or bluo-gal to detect lac gene activity in cloning procedures.
Thermo Scientific™ Pierce™ Phenol: Chloroform
Perform DNA extractions with this phenol:chloroform:isoamyl alcohol mixture (25:24:1), accompanied by a buffer to equilibrate the solution at either pH 6.7 or pH 8.0.
Thermo Scientific™ 6X Orange DNA Loading Dye Available on GSA/VA Contract for Federal Government customers only.
Prepare DNA markers and samples for loading on agarose or polyacrylamide gels withThermo Scientific™ 6X Orange Loading Dye.
Thermo Scientific™ Pierce™ Agarose
Ensure uniform lot-to-lot consistency in preparing electrophoresis gels using highly purified and blended regular- or low-melting agarose formulations.
Lonza™ AccuGENE™ 1M Tris HCl Buffers
Ready-to-use solutions ideal for a wide range of molecular biology applications
Thermo Scientific™ Pierce™ Yeast DNA Extraction Kit
Quickly, efficiently and easily lyse yeast cells to release DNA with Y-PER lysis reagent.
Thermo Scientific™ Pierce™ D-Luciferin Available on GSA/VA Contract for Federal Government customers only.
Get convenience and high performance at a low cost in firefly luciferase reporter assays with our D-Luciferin, formulated at greater than 99% purity as both monosodium and monopotassium salts.
Promega™ MagneSil™ BLUE for Wizard™ MagneSil™ Plasmid Purification System
MagneSil™ Blue for Lysate Clearing using the Wizard™ MagneSil™ Plasmid Purification System.
Thermo Scientific™ Pierce™ Glycerol
Perform a variety of protein and molecular biology applications. Thermo Scientific™ Pierce Glycerol is highly purified.
Thermo Scientific™ X-Gal Available on GSA/VA Contract for Federal Government customers only.
Effectively screen colonies with X-Gal, an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal, forming an intense blue precipitate.
Thermo Scientific™ 5-Fluoroorotic Acid
Detect expression of the URA3 gene, which encodes orotine-5'-monophosphate (OMP) dicarboxylase with Thermo Scientific™ 5-Fluoroorotic Acid, used in yeast molecular genetics.
EMD Millipore™ Calbiochem™ DNase I, Bovine Pancreas
Acts on single and double stranded DNA
EMD Millipore™ Novagen™ Pellet Paint™ Co-Precipitant Available on GSA/VA Contract for Federal Government customers only.
Compatible with most molecular biology procedures and is free of contaminating nucleic acids and nucleolytic enzymes
Thermo Scientific™ 3 M Sodium Acetate Solution, pH 5.2 Available on GSA/VA Contract for Federal Government customers only.
Precipitate DNA and RNA with this 0.22µm membrane-filtered, sterile, nuclease-free solution of sodium acetate.
Promega™ ViaFect™ Transfection Reagent
Designed to efficiently introduce DNA into a wide variety of cell lines.
Promega™ Proteinase K (PK) Solution for ReliaPrep™ Large Volume HT gDNA Isolation System
Stable at room temperature and easy to use. Proteinase K (PK) Solution is supplied at a concentration of 20mg/ml in 10mM Tris-HCl (pH 7.5), 1mM calcium chloride and 50% glycerol.
Thermo Scientific™ 2X RNA Loading Dye Available on GSA/VA Contract for Federal Government customers only.
Prepare RiboRuler RNA ladders and RNA samples for electrophoresis on agarose or polyacrylamide gels using Thermo Scientific™ 2X RNA Loading Dye.
EMD Millipore™ Novagen™ T7·Tag ™Antibody HRP Conjugate Available on GSA/VA Contract for Federal Government customers only.
Single detection reagent that eliminates need for secondary antibodies or streptavidin conjugates
EMD Millipore™ Calbiochem™ SPUTOLYSIN™ Reagent Available on GSA/VA Contract for Federal Government customers only.
For isolation of epithelial cells, pathogenic or saprophytic bacteria, fungi, and yeasts from sputum
  spinner