TMT Reagents Tandem Mass Tag
Thermo Scientific™ Pierce™ Amine-Reactive Tandem Mass Tag Reagents Available on GSA/VA Contract for Federal Government customers only.
Study protein expression by quantitative mass spectrometry using amine-labeling compounds of equal total mass (isobaric) but different reporter ions.
Thermo Scientific™ Pierce™ Amine-Reactive Tandem Mass Tag Reagents Available on GSA/VA Contract for Federal Government customers only.
For multiplex quantitative protein expression analysis by mass spectrometry
Thermo Scientific™ Immobilized Anti-TMT™ Antibody Resin
Thermo Scientific™ Pierce™ Anti-TMT Antibody Resin enables selective enrichment of peptides in complex samples that have been labeled with Tandem Mass Tag™ Reagents.
Thermo Scientific™ TMT™ Elution Buffer
Thermo Scientific™ TMT™ Elution Buffer is for use with Thermo Scientific™ Immobilized Anti-TMT™ Resin to enrich samples for peptides that have been chemically labeled with Tandem Mass Tag™ (TMT™) Reagents.
Thermo Scientific™ Pierce™ Carbonyl-reactive, 6-plex aminoxyTMT Reagents Available on GSA/VA Contract for Federal Government customers only.
Perform multiplexed characterization and quantitation of carbonyl-containing biomolecules by mass spectrometry (MS) with these carbonyl-reactive reagents.
TMT, clone: 25D5, Invitrogen™
Mouse Monoclonal Antibody
Thermo Scientific™ Pierce™ Cysteine-Reactive Tandem Mass Tag Reagents Available on GSA/VA Contract for Federal Government customers only.
Perform measurement of protein and peptide cysteine modifications (S-nitrosylation, oxidation and disulfide bridges) by multiplex quantitative mass spectrometry.
  spinner