Protein Assays and Amino Acid Analysis
BD Perm/Wash™ Buffer
Can be used in intracellular cytokine staining to permeabilize cells and as an antibody diluent and cell wash buffer
BD Cytofix/Cytoperm™ Fixation and Permeabilization Solution
1X solution can be used for the simultaneous fixation and permeabilization of cells prior to intracellular cytokine staining
BD BD GolgiStop™ Protein Transport Inhibitor (containing Monensin)
Block the intracellular protein transport processes when added to in vitro- or in vivo-stimulated lymphoid cells
Applied Biosystems™ cAMP-Screen™ Cyclic AMP Immunoassay System, 96-well Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Ultrasensitive determination of cyclic AMP (cAMP) levels in cell lysates. Thermo Scientific cAMP-Screen™ 96-well Cyclic AMP immunoassay System is a competitive chemiluminescent immunoassay formatted with maximum flexibility to permit either manual r
Promega™ FluoroTect™ GreenLys In Vitro Translation Labeling System
Allows for the fluorescent labeling and detection of proteins synthesized in vitro
Promega™ Isoquant™ Isoaspartate Detection Kit
Quantitatively detects isoaspartate resulting from aspartic acid rearrangement as well as deamidation of asparagine
Invitrogen™ CYP2B6 BACULOSOMES™ Plus Reagent, rHuman Non-distribution product as customer accommodation.
CYP2B6 BACULOSOMES™ Plus Reagent, rHuman
Invitrogen™ NA-Fluor™ Influenza Neuraminidase Assay Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
An assay for monitoring neuraminidase enzyme activity from mixed viral populations, non-viral or bacterial sources. Thermo Scientific NA-Fluor Influenza Neuraminidase Assay Kit provides validated reagents and standardized protocol for conducting neuramini
Invitrogen™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 6 Peptide Non-distribution product as customer accommodation.
Z'-LYTE Kinase Assay Kit - Ser/Thr 6 Peptide
Novex™ The FlexMAP™ 3D™ Calibration Kit Non-distribution product as customer accommodation.
The FlexMAP™ 3D™ Calibration Kit
Invitrogen™ Sapphire-II™ Luminescence Enhancer for Solution Assays Non-distribution product as customer accommodation.
Chemiluminescence enhancer that significantly reduces aqueous quenching and tune the wavelength of light emission.
Novex™ The FlexMAP™ 3D™ Performance Verification Kit Non-distribution product as customer accommodation.
The FlexMAP™ 3D™ Performance Verification Kit
Invitrogen™ I-Block™ Protein-Based Blocking Reagent Non-distribution product as customer accommodation.
Highly purified Casein-based blocking reagent. Thermo Scientific I-Block Protein-Based Blocking Reagent provides superior blocking compared to both dried milk and BSA.
R&D Systems™ E2Select Ubiquitin Conjugation Kit
Rapid and economical method to identify E2 enzyme(s) that functionally interact with an E3 ubiquitin ligase in vitro
R&D Systems™ Ubiquitin Conjugation Initiation Kit
Designed to charge ubiquitin by E1, the first reaction in the conjugation cascade
Invitrogen™ GeneBLAzer™ In Vivo Detection Kit Non-distribution product as customer accommodation.
GeneBLAzer™ In Vivo Detection Kit
Promega™ BCIP/NBT Color Development Substrate
For the colorimetric detection of alkaline phosphatase activity
Invitrogen™ NovaBright™ Phospha-Light™ EXP Assay Kit for SEAP (Secreted Placental Alkaline Phosphatase) Reporter Gene Detection Non-distribution product as customer accommodation.
Chemiluminescent reporter gene assay for the ultrasensitive detection of secreted placental alkaline phosphatase or the non-secreted placental isozyme PLAP. Thermo Scientific NovaBright Phospha-Light EXP Assay combines ultrasensitive detection with a dyna
GE Healthcare Amersham™ ECL Advance™ Blocking Reagent Available on GSA/VA Contract for Federal Government customers only.
Use for Controlling background noise.  GE Healthcare Life Sciences™ Amersham™ ECL Advance™ is Blocking Reagent controls background noise while retaining a high level of signal intensity.
BD Capture Assay Set
Measurement of cytokines levels in-vivo
R&D Systems™ Human CHIP Ubiquitin Ligase Kit, Glow-Fold Substrate
Designed for in vitro ubiquitination of user-supplied substrates by CHIP or the CHIP/HSP70/HSP40 ternary complex
Invitrogen™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 7 Peptide Non-distribution product as customer accommodation.
Z'-LYTE™ Kinase Assay Kit - Ser/Thr 7 Peptide
Invitrogen™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 18 Peptide Non-distribution product as customer accommodation.
Z'-LYTE Kinase Assay Kit - Ser/Thr 18 Peptide
Invitrogen™ Development Folders Non-distribution product as customer accommodation.
For use during imaging of blotting membranes following chemiluminescent signal development (western blotting, nucleic acid blotting).
R&D Systems™ Ubiquitin Conjugation Kit
For the formation of ubiquitinated substrate proteins
Invitrogen™ Light Emission Accelerator-II Solution Non-distribution product as customer accommodation.
Light emission triggering solution with luminescence enhancer for use with the Galacto-Light Plus™ or Dual-Light™ Reporter Gene Assay Systems.
GE Healthcare 2-D Quant Kit Available on GSA/VA Contract for Federal Government customers only.
Use for accurate determination of protein concentration in samples prepared for electrophoresis techniques. GE Healthcare Life Sciences™ 2-D Quant Kit  precipitates proteins while leaving interfering substances behind.
Promega™ cAMP, 1mM
cAMP (adenosine-3',5'-cyclic monophosphate) is an activator of cAMP-Dependent Protein Kinase, Catalytic Subunit
  spinner