Electroblotting Equipment
Invitrogen™ iBlot™ 2 Gel Horizontal Transfer Device Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Performs Western blotting transfer simply, efficiently, and reliably, within seven minutes and without the need for liquid buffers
Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems Available on GSA/VA Contract for Federal Government customers only.
Provide faster blots with higher efficiency using Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems.
Invitrogen™ iBind™ Western Device Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
An automated Western-processing device that performs every step from blocking to washes to antibody incubations via sequential lateral flow
Invitrogen™ Novex™ Bolt™ Welcome Pack + iBlot™ 2 System Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
A complete protein separation-to-western blot solution by combining a Bolt Welcome Pack A with the iBlot 2 Gel Transfer Device and iBlot 2 Transfer Stack (nitrocellulose, regular size) in lieu of one box of Bolt gels
Thermo Scientific™ Owl™ VEP-2 Mini Tank Electroblotting System Available on GSA/VA Contract for Federal Government customers only.
Produce uniform, reproducible protein transfers for up to four polyacrylamide minigels with the Thermo Scientific™ Owl™ VEP-2 Mini Tank Electroblotting System.
Invitrogen™ Mini Gel Tank and Blot Module Set Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Provides all of the necessary devices for protein separation and Western blotting using Novex™ gels, NuPAGE™ gels, and Bolt™ Bis-Tris Plus gels
Invitrogen™ Novex™ Bolt™ Vertical Mini Blot Module Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Wet transfer device used exclusively with the Bolt Minigel Tank
Hygiena ALLER-Snap Rapid Allergen Prevention Test
Liquid reagent contained in bulb of device. Pre-moistened swab in plastic tube.
Fisherbrand™ Semidry Blotting Apparatus Available on GSA/VA Contract for Federal Government customers only.
Sleek, low-profile system for fast, efficient Western transfers
EMD Millipore™ SNAP i.d.™ 2.0 Protein Detection System: Mini and Midi Blot Holders Available on GSA/VA Contract for Federal Government customers only.
Mini and midi blot holders for EMD Millipore SNAP i.d. 2.0 Protein Detection Systems
GE Healthcare Whatman™ TurboBlotter Transfer System, Large Kits Available on GSA/VA Contract for Federal Government customers only.
Downward blotting device turns the conventional Northern/Southern transfer stack upside down for rapid, high-resolution transfer of DNA and RNA
EMD Millipore™ SNAP i.d.™ 2.0 Protein Detection System: Blot Roller Available on GSA/VA Contract for Federal Government customers only.
Blot roller for EMD Millipore SNAP i.d. 2.0 Protein Detection Systems
EMD Millipore™ Luminata™ Starter Package Available on GSA/VA Contract for Federal Government customers only.
Western blot packages with unbelievable savings! Save time and money by choosing the package that best serves your needs!
EMD Millipore™ SNAP i.d. 2.0™ Protein Detection Systems for Western Blotting Available on GSA/VA Contract for Federal Government customers only.
Detection system with unique vacuum-driven technology and built-in flow distributor that actively drives reagents through membrane
EMD Millipore™ SNAP i.d.™ 2.0 Protein Detection System Available on GSA/VA Contract for Federal Government customers only.
Produces blots of a very high quality every time
EMD Millipore™ SNAP i.d.™ 2.0 Protein Detection System: Antibody Collection Tray Available on GSA/VA Contract for Federal Government customers only.
Antibody collection tray for EMD Millipore SNAP i.d. 2.0 Protein Detection Systems
EMD Millipore™ SNAP i.d.™ 2.0 Protein Detection System: Mini and Midi Blot Holding Frames Available on GSA/VA Contract for Federal Government customers only.
Mini and midi blot holding frames for EMD Millipore SNAP i.d. 2.0 Protein Detection Systems
EMD Millipore™ SNAP i.d.™ Protein Detection System: Blot Holders
Blot holders for use with EMD Millipore SNAP i.d. Protein Detection System
Invitrogen™ iBind™ Western Starter Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Complete kit for automated Western processing
GE Healthcare Whatman™ TurboBlotter Transfer System, Small Kits Available on GSA/VA Contract for Federal Government customers only.
Downward blotting device turns the conventional Northern/Southern transfer stack upside down for rapid, high-resolution transfer of DNA and RNA
GE Healthcare Accessories and Replacement Parts for Amersham™ VacuGene XL Vacuum Blotting System Available on GSA/VA Contract for Federal Government customers only.
VacuGene XL vacuum blotting system is the choice for nucleic acid blotting.
GE Healthcare Blot Module for miniVE Vertical Electrophoresis System Available on GSA/VA Contract for Federal Government customers only.
Performs 1-D and 2-D electrophoresis and electrotransfer in a single compact unit
GE Healthcare Whatman™ Minifold I Spot-Blot System, complete 96-well (acrylic) Available on GSA/VA Contract for Federal Government customers only.
Minifold I Spot-Blot system, complete 96 well (acrylic)
GE Healthcare Whatman™ Minifold I Slot-Blot System, complete 48-well (acrylic) Available on GSA/VA Contract for Federal Government customers only.
Minifold I Slot-Blot system, complete 48 well (acrylic)
GE Healthcare Whatman™ Minifold I Dot Blot System Available on GSA/VA Contract for Federal Government customers only.
Well manifolds for proteins and nucleic acids
  spinner