Electrophoresis Reagents
Thermo Scientific™ Owl™ Gel-Drying Kits 
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures with these gel-drying kits.
Thermo Scientific™ Owl™ Gel-Drying Supplies 
Use the frames and pre-cut cellophane that come in Thermo Scientific™ Owl™ Gel-Drying Supplies with Owl™ Gel-Drying Kits.
Invitrogen™ Novex™ SimplyBlue™SafeStain
Less hazardous and does not require methanol or acetic acid fixatives or destains
Thermo Scientific™ Pierce™ Silver Stain Kit 
Stain proteins in 1D and 2D gels with this rapid, ultra-sensitive and versatile silver stain system that yields consistent and reliable results.
Thermo Scientific™ Pierce™ Silver Stain Kit for Mass Spectrometry 
Reversibly stain proteins in polyacrylamide gels to identify bands to excise; then destain the gel pieces for analysis by mass spectrometry.
Thermo Scientific™ GelCode™ Blue Stain Reagent 
Achieve nanogram-level detection and results that exceed those of typical coomassie stains, all without a destaining step.
Thermo Scientific™ Pierce™ Color Silver Stain Kit
Detect proteins with this metallic silver staining technique that produces color variations to differentiate protein bands and spots in 1D and 2D gels.
Invitrogen™ Molecular Probes™ SYPRO™ Ruby Protein Gel Stain
Ideal for use in 1D and 2D PAGE
Thermo Scientific™ Pierce™ Krypton™ Fluorescent Protein Stain
Stain proteins in polyacrylamide gels or transfer membranes for fluorescent detection with popular fluorescence imagers.
Invitrogen™ Novex™ SilverXpress™ Silver Staining Kit
Staining is complete in a little over one hour
Invitrogen™ Molecular Probes™ SYPRO™ Orange Protein Gel Stain (5,000X Concentrate in DMSO)
Fast, one-step staining protocol requiring no destaining
Thermo Scientific™ GelCode™ Blue Safe Protein Stain 
Rapidly detect proteins in polyacrylamide gels with this one-step, ultrasensitive stain formulated to avoid hazardous shipping costs.
GE Healthcare PlusOne™ Silver Staining Kit for Protein 
Use for reproducible and sensitive staining of proteins in polyacrylamide gels. PlusOne™ Silver Staining Kit for Protein enables staining of proteins in less than two hours.
Invitrogen™ Novex™ Colloidal Blue Staining Kit
Five times more sensitivity than Coomassie™ R-250— and can detect as little as 10ng of BSA
Thermo Scientific™ Imperial™ Protein Stain 
Detect proteins in the low-nanogram range with this ready-to-use, in-gel, colorimetric stain formulated to deliver fast and sensitive SDS-PAGE staining.
MP Biomedicals™ Silver Enhancing Kit, 2x250mL
Used for amplifying the signal from gold particles attached to proteins or antigens immobilized on membranes
Invitrogen™ Novex™ SilverQuest™ Silver Staining Kit
30-fold more sensitive than staining with colloidal Coomassie™ G-250
Invitrogen™ Novex™ InVision™ His-Tag In-Gel Stain
One ready-to-use solution that does not require mixing, dilution, or solution preparation
Molecular Probes™ Pro-Q™ Emerald 300 Lipopolysaccharide Gel Stain Kit 
Pro-Q™ Emerald 300 Lipopolysaccharide Gel Stain Kit
Thermo Scientific™ Pierce™ Glycoprotein Staining Kit 
Rapidly stain glycosylated proteins in polyacrylamide gels using the periodic acid-Schiff (PAS) method in this convenient kit.
MilliporeSigma&trade Calbiochem™ OmniPur™ Ethidium Bromide, Molecular Biology Grade
Used for visualization of nucleic acids in gels.
Thermo Scientific™ Pierce™ Power Staining Kits
Get fast Coomassie staining of protein gels with Thermo Scientific™ Pierce™ Power Staining Kits, optimized to work exclusively with the Pierce Power Stain Cassette and Pierce Power Station.
Thermo Scientific™ Pierce™ PageBlue™ Protein Staining Solution 
A sensitive formulation of colloidal coomassie G-250 dye for endpoint staining of proteins in polyacrylamide gels and on PVDF membranes without methanol or acetic acid.
Thermo Scientific™ Pierce™ Coomassie Brilliant Blue R-250 and G-250 Dyes 
Prepare colorimetric protein gel stains using purified powdered preparations of the two most common forms of coomassie dye as the base.
Invitrogen™ Molecular Probes™ PRO-Q™ Diamond Phosphoprotein Gel Stain
Eliminates the need for blotting or phosphoprotein specific antibodies and Western blot analysis
Invitrogen™ Molecular Probes™ PRO-Q™ Diamond Phosphoprotein Enrichment Kit
Column bed volume easily scaled up or down depending on the amount of available starting material
Invitrogen™ Novex™ Lumio™ Green Detection Kit
Enables immediate visualization of Lumio™ fusion protein bands directly in a polyacrylamide gel
Invitrogen™ Novex™ SYPRO™ Ruby Protein Blot Stain
Compatible with immunostaining (Western blotting), microsequencing and mass spectrometry
Invitrogen™ Novex™ StainEase™ StainEase Staining Trays
Stains and solutions can be changed without having to handle the gel
Thermo Scientific™ Pierce™ Reversible Protein Stain Kits for Membranes
Reversibly stain proteins on nitrocellulose or PVDF membrane after transfer from polyacrylamide gels with this sensitive Ponceau S alternative.