Dye-Based Protein Gel Stains
Invitrogen™ Novex™ SimplyBlue™SafeStain Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Less hazardous and does not require methanol or acetic acid fixatives or destains
Thermo Scientific™ GelCode™ Blue Stain Reagent Available on GSA/VA Contract for Federal Government customers only.
Achieve nanogram-level detection and results that exceed those of typical coomassie stains, all without a destaining step.
Thermo Scientific™ Imperial™ Protein Stain Available on GSA/VA Contract for Federal Government customers only.
Detect proteins in the low-nanogram range with this ready-to-use, in-gel, colorimetric stain formulated to deliver fast and sensitive SDS-PAGE staining.
Thermo Scientific™ GelCode™ Blue Safe Protein Stain Available on GSA/VA Contract for Federal Government customers only.
Rapidly detect proteins in polyacrylamide gels with this one-step, ultrasensitive stain formulated to avoid hazardous shipping costs.
Invitrogen™ Novex™ Colloidal Blue Staining Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Five times more sensitivity than Coomassie™ R-250— and can detect as little as 10ng of BSA
Invitrogen™ Novex™ SilverXpress™ Silver Staining Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Staining is complete in a little over one hour
Invitrogen™ Novex™ SilverQuest™ Silver Staining Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
30-fold more sensitive than staining with colloidal Coomassie™ G-250
Invitrogen™ Novex™ InVision™ His-Tag In-Gel Stain Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
One ready-to-use solution that does not require mixing, dilution, or solution preparation
Thermo Scientific™ Pierce™ Glycoprotein Staining Kit Available on GSA/VA Contract for Federal Government customers only.
Rapidly stain glycosylated proteins in polyacrylamide gels using the periodic acid-Schiff (PAS) method in this convenient kit.
Thermo Scientific™ Pierce™ Coomassie Brilliant Blue R-250 and G-250 Dyes Available on GSA/VA Contract for Federal Government customers only.
Prepare colorimetric protein gel stains using purified powdered preparations of the two most common forms of coomassie dye as the base.
Thermo Scientific™ Pierce™ PageBlue™ Protein Staining Solution Available on GSA/VA Contract for Federal Government customers only.
A sensitive formulation of colloidal coomassie G-250 dye for endpoint staining of proteins in polyacrylamide gels and on PVDF membranes without methanol or acetic acid.
Invitrogen™ Molecular Probes™ PRO-Q™ Diamond Phosphoprotein Enrichment Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Column bed volume easily scaled up or down depending on the amount of available starting material
Invitrogen™ Molecular Probes™ PRO-Q™ Diamond Phosphoprotein Gel Stain Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Eliminates the need for blotting or phosphoprotein specific antibodies and Western blot analysis
Thermo Scientific™ Pierce™ Reversible Protein Stain Kits for Membranes
Reversibly stain proteins on nitrocellulose or PVDF membrane after transfer from polyacrylamide gels with this sensitive Ponceau S alternative.
Thermo Scientific™ Pierce™ Power Staining Kits
Get fast Coomassie staining of protein gels with Thermo Scientific™ Pierce™ Power Staining Kits, optimized to work exclusively with the Pierce Power Stain Cassette and Pierce Power Station.
Invitrogen™ Novex™ StainEase™ StainEase Staining Trays Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Stains and solutions can be changed without having to handle the gel
Invitrogen™ Novex™ SYPRO™ Ruby Protein Blot Stain Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Compatible with immunostaining (Western blotting), microsequencing and mass spectrometry
Invitrogen™ Molecular Probes™ Coomassie Fluor™ Orange Protein Gel Stain (Ready-To-Use Solution) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Staining is complete in less than an hour with no risk of overstaining the gel
Invitrogen™ Molecular Probes™ WesternDot™ 625 Goat Anti-Mouse Western Blot Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Detection performance and ease-of-use exceed existing ECL-based methods
Invitrogen™ Novex™ InVision™ His-Tag In-Gel Staining Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
One ready-to-use solution that does not require mixing, dilution, or solution preparation
Invitrogen™ Novex™ Lumio™ Green Detection Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Enables immediate visualization of Lumio™ fusion protein bands directly in a polyacrylamide gel
Invitrogen™ Molecular Probes™ PRO-Q™ Diamond Phosphoprotein Blot Stain Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Can be used to analyze the native phosphorylation state of proteins obtained from tissue specimens or from body fluids
Invitrogen™ Molecular Probes™ PRO-Q™ Diamond Phosphoprotein Gel Staining Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Can detect as little as 1-16ng of phosphoprotein per band with a linear signal over three orders of magnitude
Invitrogen™ Molecular Probes™ CandyCane™ Glycoprotein Molecular Weight Standards, Unstained Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
CandyCane glycoprotein molecular weight standards separate into alternating bands of glycosylated and nonglycosylated proteins during polyacrylamide gel electrophoresis
Fisher BioReagents™ EZ-Run™ Protein Gel Staining Solution
Ready-to-use solution for highly sensitive staining of proteins separated in polyacrylamide gels
  spinner