Protein Purification
Thermo Scientific™ 1-Step Human Coupled In Vitro Protein Expression Kits Available on GSA/VA Contract for Federal Government customers only.
Rapidly express full-length, functional proteins from mRNA or plasmid templates with yields of up to 100µg/mL per reaction using these cell-free kits.
Thermo Scientific™ Pierce™ Streptavidin Resins Available on GSA/VA Contract for Federal Government customers only.
Bind biotinylated proteins and other ligands with a wide selection of high-quality agarose and acrylamide beads and columns of recombinant streptavidin biotin-binding protein.
Pierce™ Chromogenic Endotoxin Quant Kit
A highly sensitive endpoint assay that accurately measures and detects endotoxin (lipopolysaccharide) in a protein, peptide, nucleic acids or antibody sample using the Amebocyte Lysate Assay.
Thermo Scientific™ HisPur™ Ni-NTA Resin Available on GSA/VA Contract for Federal Government customers only.
Purify His-tagged recombinant fusion proteins using high-capacity nickel-NTA (nitrilotriacetic acid) metal-chelate beads, spin columns and plates.
Thermo Scientific™ Pierce™ Detergent Removal Spin Columns and Plates Available on GSA/VA Contract for Federal Government customers only.
Rapidly eliminate high concentrations of detergents with minimal sample loss with our spin column or filter-plate formats for centrifuge processing.
Thermo Scientific™ Tween-20 Detergent Surfact-Ams™ Solution
Thermo Scientific™ Surfact-Amps 20 Detergent Solution is highly-purified Tween-20 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.
Thermo Scientific™ CarboxyLink™ Coupling Resins and Immobilization Kits
Covalently immobilize peptide antigens and other carboxylate ligands for affinity purification using DADPA-activated agarose beads and EDC crosslinker.
Invitrogen™ Total Exosome Isolation Reagent (from cell culture media) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Easily isolate exosomes and avoid time-consuming ultra-centrifugation
Invitrogen™ Molecular Probes™ NanoOrange™ Protein Quantitation Kit, 200-2,000 assays Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Sensitive and easy assay for protein quantitation, with detection as low as 10 ng/mL of protein in solution
Thermo Scientific™ GlycoLink™ Immobilization Kits and Resin
Immobilize polyclonal antibodies or other glycoproteins, via periodate-oxidized sugars, onto hydrazide-activated polyacrylamide beads for AP, IP or pull-down assays.
Thermo Scientific™ SulfoLink™ Coupling Resins and Immobilization Kits
Covalently immobilize cysteine-peptides or proteins (sulfhydryl-ligands) for affinity purification using iodoacetyl-activated agarose beads and columns.
Thermo Scientific™ Pierce™ HA Tag IP/Co-IP Kit Available on GSA/VA Contract for Federal Government customers only.
Anti-HA antibody agarose beads and complete reagent set to easily co-immunoprecipitate protein interactions involving HA-tagged fusion proteins.
Thermo Scientific™ Pierce Protease Inhibitor Tablets
Thermo Scientific™ Pierce Protease Inhibitor Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments
Thermo Scientific™ ActivX™ Fluorophosphonate Probes
Tagged phosphonate probes to assay serine hydrolase active sites using fluorescence, biotin-affinity purification or detection, or mass spectrometry.
Thermo Scientific™ Pierce™ Magnetic RNA-Protein Pull-Down Kit Available on GSA/VA Contract for Federal Government customers only.
Utilize a streamlined, robust method to enrich protein-RNA interactions using end-labeled RNA as the bait with Thermo Scientific™ Pierce Magnetic RNA-Protein Pull-Down Kit.
Thermo Scientific™ Pierce™ Streptavidin Magnetic Beads Available on GSA/VA Contract for Federal Government customers only.
Purify or immunoprecipitate biotinylated proteins or affinity ligands using streptavidin-coated magnetic particles for magnet-based separation.
Thermo Scientific™ NE-PER™ Nuclear and Cytoplasmic Extraction Kit Available on GSA/VA Contract for Federal Government customers only.
Efficiently lyse cells and extract separate cytoplasmic and nuclear protein fractions in less than two hours with this subcellular fractionation kit.
BD Recombinant Human β2 Microglobulin
Recombinant protein expressed in insect cells and purified by a combination of ion-exchange and gel-filtration methods
Thermo Scientific™ Brij™-35 Detergent Solutions
Stabilized Brij-35 detergent provided as 10% and 30% solutions for use in various protein methods including cell lysis and HPLC.
Thermo Scientific™ Pierce™ Glutathione Agarose Resin and Kits Available on GSA/VA Contract for Federal Government customers only.
Purify GST-tagged recombinant fusion proteins from cell lysates using glutathione (GSH) beaded agarose affinity purification resins, spin columns and plates.
Thermo Scientific™ Pierce™ NHS-Activated Agarose
N-Hydroxysuccinimide ester resin and columns to covalently immobilize proteins, peptides and other amine-containing ligands for affinity purification.
Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™
CAS: 151-21-3 Molecular Formula: C12H25NaO4S Molecular Weight (g/mol): 288.378 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Synonym: Sodium Lauryl Sulfate, SDS PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium;dodecyl sulfate SMILES: CCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
Thermo Scientific™ Pierce™ ConA and WGA Glycoprotein Isolation Kits
Purify glycoproteins from serum and cell lysates using affinity resins or kits with concanavilin A (ConA) or wheat germ agglutinin (WGA) lectins.
Thermo Scientific™ Halt™ Protease and Phosphatase Inhibitor Cocktails Available on GSA/VA Contract for Federal Government customers only.
Prevent protein degradation and stabilize phosphorylation with this all-in-one cocktail of protease and phosphatase inhibitors.
Thermo Scientific™ Pierce™ Protein Refolding Kit
Refold recombinant proteins that have been denatured and solubilized from inclusion bodies by determining optimal buffer conditions using this reagent matrix strategy.
Thermo Scientific™ Pierce™ HA-Tag Magnetic IP/Co-IP Kit Available on GSA/VA Contract for Federal Government customers only.
Perform immunoprecipitation (IP) assays of HA-tagged proteins or co-IP reactions with HA-tagged bait proteins with these magnetic beads, positive control and reagents.
Invitrogen™ Magnetic Stand-96 Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed for paramagnetic bead precipitation from standard 96-well, U-bottom microplates and 0.2mL PCR plates without additional accessories
Thermo Scientific™ Pierce™ Co-Immunoprecipitation Kit Available on GSA/VA Contract for Federal Government customers only.
Covalently immobilize IP antibodies and perform co-IP assays in which target protein complexes are immunoprecipitated without antibody contamination.
Thermo Scientific™ Pierce™ Magnetic ChIP Kit Available on GSA/VA Contract for Federal Government customers only.
Efficiently isolate chromatin-bound DNA by immunoprecipitation (chromatin IP) for subsequent quantitation by PCR using Thermo Scientific™ Pierce Magnetic ChIP Kit.
Thermo Scientific™ Pierce™ MS-Compatible Magnetic IP Kit (Protein A/G)
Achieve highly effective and efficient immunoprecipitation and co-immunoprecipitation of target antigens for downstream LC-MS analysis using IP antibodies and the Thermo Scientific™ Pierce™ MS-Compatible Magnetic IP Kit (Protein A/G).
  spinner