Amine Immobilization Resins and Kits
Thermo Scientific™ Pierce™ NHS-Activated Agarose
N-Hydroxysuccinimide ester resin and columns to covalently immobilize proteins, peptides and other amine-containing ligands for affinity purification.
Thermo Scientific™ AminoLink™ Plus Coupling Resin and Immobilization Kits Available on GSA/VA Contract for Federal Government customers only.
Covalently couple antibodies and other proteins via primary amines (-NH2) for affinity purification with these aldehyde-activated agarose beads.
Thermo Scientific™ AminoLink™ Coupling Resin and Immobilization Kit
Covalently immobilize proteins and antibodies by lysine residues or other primary amines (-NH2) via reductive amination with these aldehyde-activated agarose beads.
Thermo Scientific™ AminoLink™ Coupling Resin and Immobilization Kit
Easily immobilize antibodies and other proteins for use in affinity purification
Thermo Scientific™ UltraLink™ Biosupport
Covalently immobilize proteins and other primary amine biomolecules for affinity purification with this durable, azlactone-activated polyacrylamide resin.
Thermo Scientific™ Pierce™ CDI-Activated Agarose
Covalently immobilize N-nucleophiles and primary amine ligands in aqueous or organic solvent conditions using carbonyldiimidazole-activated resin.
Expedeon Amintra NHS-Activated Resin
Supplied as a suspension in isopropanol
  spinner