Biotin-Binding Affinity Resins and Beads
Thermo Scientific™ Pierce™ Streptavidin Resins Available on GSA/VA Contract for Federal Government customers only.
Bind biotinylated proteins and other ligands with a wide selection of high-quality agarose and acrylamide beads and columns of recombinant streptavidin biotin-binding protein.
Thermo Scientific™ Pierce™ NeutrAvidin™ Resins Available on GSA/VA Contract for Federal Government customers only.
Acrylamide and agarose beads and columns of specially-modified, pH-stabilized avidin; the low-cost alternative to streptavidin biotin-binding supports.
GE Healthcare Streptavidin Sepharose™ High Performance Medium Available on GSA/VA Contract for Federal Government customers only.
Used for convenient, fast, reliable purification of biotinylated compounds. Streptavidin Sepharose™ High Performance Medium is a physically and chemically stable medium.
Thermo Scientific™ Pierce™ Monomeric Avidin Resins Available on GSA/VA Contract for Federal Government customers only.
Reversibly bind biotinylated proteins and other ligands with monomers of avidin covalently attached as individual subunits onto beaded resins.
Thermo Scientific™ Pierce™ Avidin Agarose Available on GSA/VA Contract for Federal Government customers only.
Avidin biotin-binding protein immobilized onto beaded agarose resin for affinity purification methods involving biotinylated proteins and ligands.
GE Healthcare Streptavidin HP SpinTrap™ Kit Available on GSA/VA Contract for Federal Government customers only.
Prepacked single-use spin columns for simple, small-scale protein enrichment through immobilized biotinylated biomolecules
EMD Millipore™ Upstate Streptavidin, Agarose Conjugate Available on GSA/VA Contract for Federal Government customers only.
Used for purification of biotinylated macromolecules.
EMD Millipore™ Novagen™ Streptavidin Agarose Available on GSA/VA Contract for Federal Government customers only.
Consists of cross-linked agarose covalently coupled with pure streptavidin.
Promega High Capacity Magne Streptavidin Beads, 3ml
High Capacity Magne Streptavidin Beads are magnetic affinity beads with high specificity and high capacity for binding biotinylated antibodies and proteins. Goat Anti-Human Biotinylated IgG is 0.5mg/ml in PBS (pH 7.4) with 0.1% sodium azide.
Promega Goat Anti-Human Biotinylated IgG, 4ml
High Capacity Magne Streptavidin Beads are magnetic affinity beads with high specificity and high capacity for binding biotinylated antibodies and proteins. Goat Anti-Human Biotinylated IgG is 0.5mg/ml in PBS (pH 7.4) with 0.1% sodium azide.
Promega SoftLink™ Soft Release Avidin Resin
Provides reversible binding of biotinylated proteins under mild, nondenaturing conditions
GE Healthcare GST SpinTrap™ Available on GSA/VA Contract for Federal Government customers only.
For rapid small-scale purification of GST-tagged proteins directly from clarified or unclarified cell lysates
GE Healthcare HiScreen Capto™ L Columns Available on GSA/VA Contract for Federal Government customers only.
HiScreen Capto L columns are prepacked with Capto L, an affinity chromatography medium (resin) for capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs.
GE Healthcare Streptavidin HP MultiTrap™ Plates Available on GSA/VA Contract for Federal Government customers only.
Fast, reliable protein enrichment through immobilized biotinylated biomolecules in prepacked 96-well filter plates
Expedeon Amintra Streptavidin Resin
Offers an ultra-high biotin-binding capacity with a very low leaching
  spinner