Carbohydrate Immobilization Resins and Kits
Thermo Scientific™ GlycoLink™ Immobilization Kits and Resin
Immobilize polyclonal antibodies or other glycoproteins, via periodate-oxidized sugars, onto hydrazide-activated polyacrylamide beads for AP, IP or pull-down assays.
  spinner