Carboxyl Immobilization Resins and Kits
Thermo Scientific™ CarboxyLink™ Coupling Resins and Immobilization Kits
Covalently immobilize peptide antigens and other carboxylate ligands for affinity purification using DADPA-activated agarose beads and EDC crosslinker.
  spinner