Detergent Removal Resins and Kits
Thermo Scientific™ Pierce™ Detergent Removal Spin Columns and Plates Available on GSA/VA Contract for Federal Government customers only.
Rapidly eliminate high concentrations of detergents with minimal sample loss with our spin column or filter-plate formats for centrifuge processing.
Thermo Scientific™ HiPPR Detergent Removal Resin and Kits Available on GSA/VA Contract for Federal Government customers only.
Efficiently remove detergents from samples with this high protein and peptide recovery detergent removal resin, especially for dilute protein samples.
Thermo Scientific™ Pierce™ SDS-Out™ SDS Precipitation Reagent
Quickly remove SDS from small-volume (0.1 to 10mL) protein samples with this SDS precipitation kit.
EMD Millipore™ Calbiochem™ Reagent Grade TCEP, Hydrochloride Reducing Agent Available on GSA/VA Contract for Federal Government customers only.
Water-soluble and odorless disulfide-reducing agent that is more stable and effective than DTT
  spinner