Detergents for Cell Lysis
Thermo Scientific™ Tween-20 Detergent Surfact-Ams™ Solution
Thermo Scientific™ Surfact-Amps 20 Detergent Solution is highly-purified Tween-20 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.
Thermo Scientific™ Brij™-35 Detergent Solutions
Stabilized Brij-35 detergent provided as 10% and 30% solutions for use in various protein methods including cell lysis and HPLC.
Thermo Scientific™ NP-40 Detergent Surfact-Ams™ Solution Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Surfact-Amps NP-40 Detergent Solution is highly-purified NP-40 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.
Thermo Scientific™ Tween-80 Detergent Surfact-Ams™ Solution
Thermo Scientific™ Surfact-Amps 80 Detergent Solution is highly-purified Tween-80 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.
Thermo Scientific™ Triton X-100 Detergent Surfact-Ams™ Solution Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Surfact-Amps X-100 Detergent Solution is highly-purified Triton X-100 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.
Thermo Scientific™ Pierce™ CHAPS Available on GSA/VA Contract for Federal Government customers only.
Non-denaturing Zwitterionic detergents that disrupt nonspecific protein interactions with less protein aggregation than nonionic detergents.
Thermo Scientific™ Pierce™ Sodium Deoxycholate
Water-soluble, ionic detergent used in cell lysis buffers (e.g., RIPA buffer) to disrupt protein interactions for liposome and membrane protein isolation.
Thermo Scientific™ Pierce™ Octylthioglucoside (OTG)
Lyse cells and solubilize proteins without denaturing them using this low-MW nonionic detergent that is resistant to beta-D-glucoside digestion (unlike Octyl-beta-glucoside).
Promega™ Digitonin Detergent Solution
Detergent solution for permeabilizing cells.
Thermo Scientific™ Pierce™ Sodium Dodecyl Sulfate (SDS) C12
Highly purified form of SDS especially suited for protein experiments requiring tightly controlled solubility parameters.
Thermo Scientific™ Pierce™ Sodium Dodecyl Sulfate (SDS) Lauryl
Perform routine protein electrophoresis and cell lysis methods using this popular, standard-grade SDS detergent.
Thermo Scientific™ Pierce™ Octyl beta-Glucoside
Nonionic detergent widely used for membrane protein solubilization that is dialyzable but susceptible to beta-Glucosidase digestion (unlike OTG).
Thermo Scientific™ Triton X-114 Detergent Surfact-Amps™ Solution
Purified nonionic Triton X-114 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.
Thermo Scientific™ Pierce™ n-Dodecyl-β-Maltoside Available on GSA/VA Contract for Federal Government customers only.
Water-soluble, non-denaturing and nonionic detergent especially useful for solubilizing hydrophobic membrane proteins to preserve their activity.
Thermo Scientific™ Brij™-58 Detergent Solution
Purified Brij-58 detergent that is stabilized and supplied as a 10% solution in 10mL glass ampules for protein research and molecular biology.
Thermo Scientific™ Surfact-Amps™ Purified Detergent Sampler
Convenient collection of various Surfact-Amps Detergent Solutions and other selected useful detergents for protein research methods.
MilliporeSigma&trade Novagen™ PopCulture™ Reagent Available on GSA/VA Contract for Federal Government customers only.
Formulated to extract proteins from E. coli cells directly in culture medium.
  spinner