Enzyme Active-Site Probes and Enrichment Kits
Thermo Scientific™ Pierce™ GTPase Enrichment Kit and Probes
A desthiobiotin-GTP probe kit to label, affinity purify and measure the active site of GTPases and the GTPase subunits of G-protein coupled receptors.
Thermo Scientific™ ActivX™ Fluorophosphonate Probes
Tagged phosphonate probes to assay serine hydrolase active sites using fluorescence, biotin-affinity purification or detection, or mass spectrometry.
Thermo Scientific™ Pierce™ Kinase Enrichment Kits and Probes
Desthiobiotin-ATP and -ADP probes to label, affinity purify and measure the active site of ATPases (kinases), including chaperones and metabolic enzymes.
  spinner