Gel Filtration Media and Columns
Cytiva (Formerly GE Healthcare Life Sciences) Sephadex™ LH-20 Available on GSA/VA Contract for Federal Government customers only.
Use for molecular sizing of natural product such as steroid, terpenoid, lipid, low molecular weight peptide. Cytiva Life Sciences™Sephadex™ LH-20 is a liquid chromatography media useful to analytical and industrial scale for preparation of closely related molecular species.
Cytiva (Formerly GE Healthcare Life Sciences) Superdex™ 200 Increase 10/300 GL Prepacked Tricorn™ Columns Available on GSA/VA Contract for Federal Government customers only.
For size exclusion chromatography and analysis of proteins with molecular weights between 10,000 and 600,000. Cytiva Lifescience™ Superdex™ 200 Increase 10/300 GL Prepacked Tricorn™ Columns are designed for small scale preparative purification.
Cytiva (Formerly GE Healthcare Life Sciences) Gel Filtration Calibration Kits Available on GSA/VA Contract for Federal Government customers only.
Calibrate gel filtration columns simply, reliably. Cytiva Life Sciences™ Gel Filtration Calibration Kits contain a well-defined series of protein standards.
Cytiva (Formerly GE Healthcare Life Sciences) HiTrap™ Heparin HP Columns Available on GSA/VA Contract for Federal Government customers only.
Purify DNA-binding proteins, coagulation factors, lipoproteins and protein synthesis factors. Cytiva Life Sciences™ HiTrap™ Heparin HP Columns are prepacked with Heparin Sepharose™ High Performance for high-resolution purification.
Cytiva (Formerly GE Healthcare Life Sciences) Sephadex™ Ion Exchange Media Available on GSA/VA Contract for Federal Government customers only.
Separate molecules over broad molecular weight and pH ranges
Cytiva (Formerly GE Healthcare Life Sciences) Sephadex™ G50 Superfine Media
Use as a well established gel filtration medium for desalting and buffer exchange of biomolecules >30 000 molecular weight. Sephadex™ G50 Superfine Media's small bead size gives higher efficiency.
Cytiva (Formerly GE Healthcare Life Sciences) Sepharose Agarose Gel Filtration Media Available on GSA/VA Contract for Federal Government customers only.
Choose from a wide range of bead-formed agarose-based gel filtration media. Cytiva Life Sciences™ Sepharose Agarose Gel Filtration Media shows good separation over a wide molecular weight range.
Cytiva (Formerly GE Healthcare Life Sciences) Superdex™ 75 Prep Grade
Offers high resolution fractionation for separation, polishing and product formulation of recombinant proteins
Cytiva (Formerly GE Healthcare Life Sciences) Superdex™ 200 Prep Grade Available on GSA/VA Contract for Federal Government customers only.
Produce separation medium with high selectivity and resolution.  Cytiva Life Sciences™ Superdex™ 200 Prep Grade for purifying proteins of fractionation range: 10000 to 600000D
Cytiva (Formerly GE Healthcare Life Sciences) Superdex™ 200 Increase 5/150 GL Prepacked Tricorn™ Columns
For size exclusion chromatography and analysis of proteins with molecular weights between 10,000 and 600,000.  Cytiva Lifescience™ Superdex™ 200 Increase 5/150 GL Prepacked Tricorn™ Columns are designed for small scale preparative purification.
Cytiva (Formerly GE Healthcare Life Sciences) Sephadex™ G-25 Media Available on GSA/VA Contract for Federal Government customers only.
Use of Sephadex™ is  well established as a gel filtration medium for desalting and buffer exchange in industrial applications. Cytiva Life Sciences™ Sephadex™ G-25 Media quickly desalts, removes contaminants and transfers to a new buffer in a single step.
Cytiva (Formerly GE Healthcare Life Sciences) Superose™ Media Prep Grade Lab Packs
High performance gel filtration media for scaleup to preparative purifications of biomolecules
Cytiva (Formerly GE Healthcare Life Sciences) Sephadex™ G15 Media
Gel filtration medium for desalting and buffer exchange for small biomolecules greater than 1500 molecular weight
Cytiva (Formerly GE Healthcare Life Sciences) IgG Sepharose™ 6 Fast Flow Medium Available on GSA/VA Contract for Federal Government customers only.
Use for rapid purification of protein A and protein A fusion conjugates. IgG Sepharose™ 6 Fast Flow Medium achieves the highest flowrates.
Cytiva (Formerly GE Healthcare Life Sciences) Sephacryl™ S-100 High Resolution Media
Hydrophilic, rigid allyl dextran/bisacrylamide matrix for fast and reproducible purification of proteins, polysaccharides and other macromolecules
Cytiva (Formerly GE Healthcare Life Sciences) HiTrap™ Benzamidine FF (high sub) Columns
Remove or purify serine proteases such as thrombin. Cytiva Life Sciences™ HiTrap™ Benzamidine FF (high sub) Columns are prepacked with Benzamidine Sepharose 4 Fast Flow (high sub).
Cytiva (Formerly GE Healthcare Life Sciences) HiTrap™ NHS-Activated HP Columns Available on GSA/VA Contract for Federal Government customers only.
Couple ligands containing primary amines easily. Cytiva Life Sciences™ HiTrap™ NHS-Activated HP Columns are prepacked with NHS activated Sepharose™ High Performance.
Cytiva (Formerly GE Healthcare Life Sciences) HiPrep™ Sephacryl™ HR Prepacked Gel Filtration Columns Available on GSA/VA Contract for Federal Government customers only.
For reproducible protein purification, separating polysaccarides, macromolecules with extended structures and small plasmids
Cytiva (Formerly GE Healthcare Life Sciences) Sephacryl™ S-200 High Resolution Media Available on GSA/VA Contract for Federal Government customers only.
Hydrophilic, rigid allyl dextran/bisacrylamide matrix for fast and reproducible purification of proteins, polysaccharides and other macromolecules
Cytiva (Formerly GE Healthcare Life Sciences) illustra™ Sephadex™ G-50 and G-100 DNA Grade Agarose Gels Available on GSA/VA Contract for Federal Government customers only.
Used for purification of DNA from small molecules.  Cytiva illustra™ Sephadex™ G-50 and G-100 DNA Grade Agarose Gels purify DNA by gel filtration.
Cytiva (Formerly GE Healthcare Life Sciences) Sephadex™ G-100 Media
Commonly used for desalting and buffer exchange of very large biomolecules
Cytiva (Formerly GE Healthcare Life Sciences) HiTrap™ Blue HP Columns
For purifying albumin, enzymes including NAD+ and NADP+, coagulation factors, interferons and related proteins
Cytiva (Formerly GE Healthcare Life Sciences) Sephadex™ G10 Media
Gel filtration medium for desalting and buffer exchange of peptides and small biomolecules
R&D Systems™ Proteome Purify 2 Mouse Serum Protein Immunodepletion Resin Available on GSA/VA Contract for Federal Government customers only.
Provides an easy-to-use, high capacity system to reproducibly remove albumin and IgG from mouse serum or plasma
  spinner