GST-tagged Protein Purification Resins and Kits
Thermo Scientific™ Pierce™ Glutathione Agarose Resin and Kits Available on GSA/VA Contract for Federal Government customers only.
Purify GST-tagged recombinant fusion proteins from cell lysates using glutathione (GSH) beaded agarose affinity purification resins, spin columns and plates.
Thermo Scientific™ Pierce™ Glutathione Superflow Agarose
Perform high-purity, large-scale purification of GST-tagged proteins with glutathione that has been immobilized to a crosslinked 6% beaded agarose.
GE Healthcare Glutathione Sepharose™ 4 Fast Flow Media Available on GSA/VA Contract for Federal Government customers only.
Single-step purification of GST-tagged proteins and other glutathione S-transferase or glutathione-dependent proteins. Glutathione Sepharose 4 Fast Flow Media is used for the purification of GST-tagged proteins from bacterial lysates and other S-tranferases or glutathione-dependent proteins.
GE Healthcare GSTrap™ HP Columns Available on GSA/VA Contract for Federal Government customers only.
Use to provide high-resolution, one-step purifications of glutathione S-Transferase (GST) tagged proteins. GE Healthcare Life Sciences™ GSTrap™ HP Columns are prepacked with Glutathione Sepharose™ High Performance medium.
Thermo Scientific™ Pierce™ GST (Glutathione S-Transferase)
Pure, lyophilized glutathione S-transferase (GST) protein from Schistosoma japonicum for use in recombinant fusion protein research methods and assays.
GE Healthcare Glutathione Sepharose™ High Performance Media Available on GSA/VA Contract for Federal Government customers only.
For high resolution purification of Glutathione S transferase (GST) tagged proteins
GE Healthcare GST Gene Fusion System pGEX Vectors Available on GSA/VA Contract for Federal Government customers only.
Designed for inducible, high-level intracellular expression of genes or gene fragments as fusions with Schistosoma japonicum GST
GE Healthcare GSTrap™ FF Columns: 16 × 25mm Available on GSA/VA Contract for Federal Government customers only.
Purifies GST-tagged proteins in one step
EMD Millipore™ Novagen™ His Bind™ Resin Available on GSA/VA Contract for Federal Government customers only.
Suited to small to medium scale purifications.
EMD Millipore™ Novagen™ GST Bind™ Resin Available on GSA/VA Contract for Federal Government customers only.
Utilizes 14-atom spacer arm to covalently attach reduced glutathione via sulfide linkage.
GE Healthcare GSTrap™ HP Columns PROMO
Use to provide high-resolution, one-step purifications of glutathione S-Transferase (GST) tagged proteins. GE Healthcare Life Sciences™ GSTrap™ HP Columns are prepacked with Glutathione Sepharose™ High Performance medium.
GE Healthcare pGEX 3' Sequencing Primer, 5'-d[CCG-GGAGCTGCATGTGTCAGAGG]-3 Available on GSA/VA Contract for Federal Government customers only.
Ready for immediate use. GE Healthcare pGEX 3' Sequencing Primer, 5'-d[CCG-GGAGCTGCATGTGTCAGAGG]-3 is a vector primer for sequencing double-stranded DNA inserted into pGEX vectors using the Glutathione S-transferase (GST) Gene Fusion System.
GE Healthcare GST Bulk Kit Available on GSA/VA Contract for Federal Government customers only.
For small-scale manual purification of GST-tagged proteins using either column chromatography or batch method. Facilitates optimization studies of expression, solubility conditions and purification parameters of GST-tagged proteins
GE Healthcare GSTrap™ 4B Prepacked Columns: 16 × 25mm Available on GSA/VA Contract for Federal Government customers only.
For fast and simple purification of GST-tagged proteins
GE Healthcare GSTrap™ FF Columns: 7 × 25mm Available on GSA/VA Contract for Federal Government customers only.
Purifies GST-tagged proteins in one step
GE Healthcare GST MultiTrap™ 4B Plates Available on GSA/VA Contract for Federal Government customers only.
96-well filter plates for high-throughput screening and parallel purification of GST-tagged proteins
GE Healthcare GSTrap™ 4B Prepacked Columns: 7 × 25mm Available on GSA/VA Contract for Federal Government customers only.
Use for convenient, high capacity one-step purification of glutathione S-transferase (GST) tagged proteins. GE Healthcare Life Sciences™ GSTrap™ 4B Prepacked Columns: 7 × 25mm are prepacked with Glutathione Sepharose™ 4B
EMD Millipore™ Calbiochem™ Heparin Binding Epidermal Growth Factor, Human
Receptor for diphtheria toxin and functions in juxtacrine signaling in cells.
Expedeon Amintra GST-Tagged Magnetic Resin
For magnetic purification of proteins with high process flexibility and higher protein yield at shortened purification time
GE Healthcare GSTrap™ FF Columns: 7 × 25mm: PROMOTION
Use these pre-packed Glutathione Sepharose Fast Flow columns for fast, convenient, one-step purification of glutathione S-transferase (GST) tagged proteins. GE Healthcare HiPrep™ GSTrap™ FF Columns allow easy scale-up by connecting the columns in series.
GE Healthcare GSTrap™ FF Columns: 16 × 25mm : PROMOTION
Promotion Details: Receive a 15&discount.
  spinner