In vitro Protein Expression Systems
Thermo Scientific™ 1-Step Human Coupled In Vitro Protein Expression Kits Available on GSA/VA Contract for Federal Government customers only.
Rapidly express full-length, functional proteins from mRNA or plasmid templates with yields of up to 100µg/mL per reaction using these cell-free kits.
Promega™ E. coli S30 T7 High-Yield Protein Expression System
Express up to 500μg/mL of protein in 1 hour from plasmid vectors containing a T7 promoter and a ribosome binding site
Thermo Scientific™ 1-Step Human High-Yield In Vitro Expression Kits Available on GSA/VA Contract for Federal Government customers only.
Express functional proteins with 10 to 100 times greater yield per mL than other mammalian IVT systems with Thermo Scientific™ 1-Step Human High-Yield In Vitro Translation (IVT) Kits.
Promega™ TNT™ SP6 High-Yield Protein Expression System
A single-tube, coupled transcription/translation system designed to express proteins in only two hours
EMD Millipore™ Novagen™ pET-32a(+) DNA Available on GSA/VA Contract for Federal Government customers only.
Designed for cloning high-level expression of peptide sequences.
Thermo Scientific™ T7 Cell-Free Expression Vectors
Generate His-, HA-, Myc- or other label-tagged fusion proteins by cell-free in vitro translation (IVT) with these versatile T7 expression vectors.
Promega™ pH6HTN His6HaloTag™ T7 Vector for E. coli and Cell-Free Protein Expression
For inducible expression of HaloTag fusion proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter
Promega™ pH6HTC His6HaloTag™ T7 Vector for E. coli and Cell-Free Protein Expression
For inducible expression of HaloTag fusion proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter
Promega™ TNT™ T7 Insect Cell Extract Protein Expression System
A convenient, quick, single-tube, coupled transcription/translation system for the cell-free expression of proteins
Gibco™ Expi293™ Expression System Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed with transient expression technology for rapid and ultra high-yield protein production from mammalian cells. Gibco™ Expi293™ Expression System Kit is based on high density culture of Expi293F™ cells in Expi293™ Expression Medium.
Promega™ Novobiocin Sodium Salt
Modulate protein expression in mammalian cells.
Thermo Scientific™ 1-Step Heavy Protein IVT Kit
Express stable isotope-labeled (i.e., heavy) proteins by in vitro translation (IVT) for mass spectrometry using these cell-free expression kits.
EMD Millipore™ Novagen™ pET-32b(+) DNA Available on GSA/VA Contract for Federal Government customers only.
Designed for cloning high-level expression of peptide sequences.
EMD Millipore™ Novagen™ pET-32c(+) DNA
Designed for cloning high-level expression of peptide sequences.
EMD Millipore™ Novagen™ pET-32 Ek/LIC Vector Kit
Designed for cloning high-level expression of target proteins.
Promega™ HaloLink™ Resins
Solid support that allows covalent and oriented attachment of HaloTag™ fusion proteins to a Sepharose™ surface
Promega™ pFN29A His6HaloTag™ T7 Flexi T7 Vector for E. coli and Cell-Free Protein Expression
For inducible expression of HaloTag fusion proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter
Promega™ pFN29K His6HaloTag™ T7 Flexi Vector for E. coli and Cell-Free Protein Expression
For inducible expression of HaloTag fusion proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter
Promega™ pFC30A His6HaloTag™ T7 Flexi Vector for E. coli and Cell-Free Protein Expression
For inducible expression of HaloTag fusion proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter
Promega™ pFC30K His6HaloTag™ T7 Flexi Vector for E. coli and Cell-Free Protein Expression
For inducible expression of HaloTag fusion proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter
Invitrogen™ BacMam LRRK2-GFP Wild Type Reagent Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
BacMam LRRK2-GFP Wild Type Reagent
Invitrogen™ BacMam LRRK2-GFP G2019S Reagent Non-distribution product as customer accommodation.
BacMam LRRK2-GFP G2019S Reagent
  spinner