Misc Immobilization Supports
Thermo Scientific™ Pierce™ IgG Plus Orientation Kits Available on GSA/VA Contract for Federal Government customers only.
Bind and covalently immobilize antibodies to create a reusable affinity purification column with high binding capacity with Protein A or G agarose columns and DSS crosslinker.
  spinner