Protease Inhibitors and Phosphatase Inhibitors
Thermo Scientific™ Pierce Protease Inhibitor Tablets
Thermo Scientific™ Pierce Protease Inhibitor Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments
Thermo Scientific™ Halt™ Protease and Phosphatase Inhibitor Cocktails Available on GSA/VA Contract for Federal Government customers only.
Prevent protein degradation and stabilize phosphorylation with this all-in-one cocktail of protease and phosphatase inhibitors.
Thermo Scientific™ Halt™ Protease Inhibitor Cocktails Available on GSA/VA Contract for Federal Government customers only.
Inhibit proteolytic degradation during cell lysis and protein extraction with these ready-to-use concentrated stock solutions of protease inhibitors.
Thermo Scientific™ Pierce Protease Inhibitor Mini Tablets
Thermo Scientific™ Pierce Protease Inhibitor Mini Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments
Thermo Scientific™ Halt™ Phosphatase Inhibitor Cocktail Available on GSA/VA Contract for Federal Government customers only.
Stabilize serine, threonine and tyrosine phosphorylation of proteins in cells and tissues with this cocktail of phosphatase inhibitors.
Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets
Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets broad-spectrum protease and pghospahatase inhibitor formulation provides complete protection from dephosphorylation and proteolytic degradation.
Invitrogen™ Novex™ AcTEV™ Protease Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Specifically recognizes a seven amino acid sequence (Glu-Asn-Leu-Tyr-Phe-Gln-Gly, cleaving between Gln and Gly), ideal for removing affinity tags from fusion proteins
EMD Millipore™ Calbiochem™ N-Ethylmaleimide Available on GSA/VA Contract for Federal Government customers only.
Sulfhydryl alkylating reagent that inhibits H+-ATPase and suppresses the short circuit current (IC50 = 22 μM) in pancreatic duct cells.
Thermo Scientific™ Pierce Phosphatase Inhibitor Mini Tablets
Thermo Scientific™ Pierce Phosphatase Inhibitor Mini Tablets provide complete cell lysate and biological sample protection from dephosphorylation with a broad spectrum phosphatase inhibitor formulation.
Thermo Scientific™ Pierce™ Protease Inhibitor Tablets
Thermo Scientific™ Pierce Protease Inhibitor Tablets and Mini Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments.
Thermo Scientific™ Protease Inhibitor Cocktails Available on GSA/VA Contract for Federal Government customers only.
Protect your sample from proteolytic degradation during cell lysis with these broad-spectrum protease inhibitor cocktails.
Invitrogen™ Novex™ SUMO Protease Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
A recombinant fragment of ULP1 (Ubl-specific protease 1) from Saccharomyces cerevisiae
EMD Millipore™ Calbiochem™ Protease Inhibitor Cocktail Set III, EDTA-Free Available on GSA/VA Contract for Federal Government customers only.
Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.
EMD Millipore™ Calbiochem™ Protease Inhibitor Cocktail Set V, EDTA-Free Available on GSA/VA Contract for Federal Government customers only.
A cocktail of four protease inhibitors for the inhibition of serine, cysteine, but not metalloproteases.
EMD Millipore™ Calbiochem™ Smoothened Agonist, SAG Available on GSA/VA Contract for Federal Government customers only.
A cell-permeable Smoothened Agonist, SAG, CAS 364590-63-6, modulates the coupling of Smo with its downstream effector by interacting with the Smo heptahelical domain (KD = 59 nM).
EMD Millipore™ Calbiochem™ Protease Inhibitor Cocktail Set I Available on GSA/VA Contract for Federal Government customers only.
A cocktail of five protease inhibitors that will inhibit a broad range of proteases and esterases. Supplied with a data sheet.
EMD Millipore™ Calbiochem™ Fucosyltransferase Inhibitor, 2F-Peracetyl-Fucose
2F-Peracetyl-Fucose is a cell-permeable fluorinated fucose derivative that acts as an inhibitor of fucosyltransferases following its uptake and metabolic transformation into a GDP-fucose mimetic.
Thermo Scientific™ Pierce™ Protease Inhibitors Available on GSA/VA Contract for Federal Government customers only.
Customize your protease inhibitor cocktail formulation with these small packages of individual protease inhibitor peptides and compounds.
EMD Millipore™ Calbiochem™ Phosphatase Inhibitor Cocktail Set II Available on GSA/VA Contract for Federal Government customers only.
A cocktail of five phosphatase inhibitors for the inhibition of acid and alkaline phosphatases as well as protein tyrosine phosphatases (PTPs). Suitable for use with cell lysates and tissue extracts.
EMD Millipore™ Calbiochem™ InSolution™ Y-27632
Y-27632A, CAS 146986-50-7, is a cell-permeable, reversible, inhibitor of Rho kinases (Ki = 140 nM for p160ROCK). Enhances survival & cloning efficiency of ESC without affecting their pluripotency.
Promega™ Protease Inhibitor Cocktail
A mixture of six different protease inhibitors with different target protease specificities
EMD Millipore™ Calbiochem™ Cathepsin D/E Substrate, Fluorogenic
An internally quenched fluorogenic 11-mer peptide that acts as a sensitive and selective substrate for cathepsins D and E, but not cathepsins B, H, or L .
EMD Millipore™ Calbiochem™ InSolution™ TAPI-2 Available on GSA/VA Contract for Federal Government customers only.
A 10 mM (1 mg/241 μL) sterile-filtered solution of TAPI-2 in H2O.
EMD Millipore™ Calbiochem™ Rho Kinase Inhibitor Available on GSA/VA Contract for Federal Government customers only.
The Rho kinase inhibitor, CAS 872543-07-6, is a cell-permeable, highly specific, reversible, potent, and ATP-competitive inhibitor of Rho-associated kinase (ROCK; Ki = 1.6 nM).
EMD Millipore™ Calbiochem™ Adenosine 3′,5′-cyclic Monophosphate, N6,O2′-Dibutyryl-, Sodium Salt Available on GSA/VA Contract for Federal Government customers only.
This Dibutyryl-cAMP, CAS 16980-89-5, is a cell-permeable cAMP analog that preferentially activates PKA.
EMD Millipore™ Calbiochem™ InSolution™ Rapamycin Available on GSA/VA Contract for Federal Government customers only.
InSolution™ Rapamycin, CAS 53123-88-9, is a 5 mM (500 μg/109 μl) solution of Rapamycin (Cat. No. 553210) in DMSO. Selectively inhibits mTOR and blocks activation of p70 S6 Kinase (IC50 = 50 pM).
EMD Millipore™ Calbiochem™ Latrunculin A, Latrunculia magnifica Available on GSA/VA Contract for Federal Government customers only.
Latrunculin A, CAS 76343-93-6, is a cell-permeable marine toxin that disrupts microfilament organization in cultured cells by the formation of a 1:1 complex with monomeric G-actin (KD = 200 nM).
Acros Organics™ Lopinavir, 98%
CAS: 192725-17-0 Molecular Formula: C37H48N4O5 Molecular Weight (g/mol): 628.814 InChI Key: KJHKTHWMRKYKJE-PYJUJTJGSA-N PubChem CID: 133109001 IUPAC Name: N-[(2R,4R,5R)-5-[[2-(2,6-dimethylphenoxy)acetyl]amino]-4-hydroxy-1,6-diphenylhexan-2-yl]-3-methyl-2-(2-oxo-1,3-diazinan-1-yl)butanamide SMILES: CC1=C(C(=CC=C1)C)OCC(=O)NC(CC2=CC=CC=C2)C(CC(CC3=CC=CC=C3)NC(=O)C(C(C)C)N4CCCNC4=O)O
Thermo Scientific™ Pepstatin A
Protease inhibitor, inhibits Pepsin, Renin and Cathepsin D.
  spinner