Protein Affinity Purification Reagents and Kits
BD Recombinant Human β2 Microglobulin Available on GSA/VA Contract for Federal Government customers only.
Recombinant protein expressed in insect cells and purified by a combination of ion-exchange and gel-filtration methods
eBioscience™ Protein Transport Inhibitor Cocktail, 500x Non-distribution product as customer accommodation.
Addition to cells in culture inhibits intracellular protein secretory/transport pathway, resulting in accumulation of secreted proteins in the lumen of the ER and in the Golgi apparatus.
Novus Biologicals™ Protein Thiolation Kit
Designed to facilitate the conjugation of proteins to maleimide-activated labels
R&D Systems™ Universal Kinase Activity Kit Available on GSA/VA Contract for Federal Government customers only.
A simple, nonradioactive high throughput-compatible format for assaying the enzyme activity of kinases in vitro
EMD Millipore™ Novagen™ Thrombin Cleavage Capture Kit Available on GSA/VA Contract for Federal Government customers only.
Designed for cleavage of fusion proteins.
Promega™ HaloTag™ Protein Purification System
Purifies proteins fused to HaloTag, a novel protein tag that enhances the expression and solubility of recombinant proteins
R&D Systems™ Sialyltransferase Activity Kit Available on GSA/VA Contract for Federal Government customers only.
A simple, nonradioactive high throughput-compatible format for assaying the enzyme activity of all sialyltransferases regardless of different acceptor substrates
EMD Millipore™ Novagen™ Factor Xa Cleavage Capture Kit Available on GSA/VA Contract for Federal Government customers only.
Designed for specific cleavage of fusion proteins.
EMD Millipore™ Novagen™ Enterokinase Cleavage Capture Kit Available on GSA/VA Contract for Federal Government customers only.
Designed for specific cleavage of fusion proteins.
Novus Biologicals™ Protein A - Agarose
Protein A - Agarose (IgGBind) for Immunoprecipitation
EMD Millipore™ Novagen™ Recombinant Enterokinase Available on GSA/VA Contract for Federal Government customers only.
Purified preparation of catalytic subunit of bovine enterokinase.
EMD Millipore™ Novagen™ Xarrest™ Agarose Available on GSA/VA Contract for Federal Government customers only.
Removes target proteins by affinity capture
Novus Biologicals™ Control Resin
Control resin support product for research applications
Novus Biologicals™ Maleimide Activated Ovalbumin
For conjugation of Ovalbumin to proteins, peptides and ligands that contain sulfhydryl (-SH) groups
R&D Systems™ Universal Sulfotransferase Activity Kit Available on GSA/VA Contract for Federal Government customers only.
A simple, nonradioactive high throughput-compatible format for assaying the enzyme activity of all sulfotransferases that use 3-phosphoadenosine-5-phosphosulfate (PAPS) as the donor substrate
Novus Biologicals™ Maleimide Activated Streptavidin
For conjugation of Streptavidin to proteins, peptides and ligands that contain sulfhydryl (-SH) groups
R&D Systems™ Glycosyltransferase Activity Kit Available on GSA/VA Contract for Federal Government customers only.
A simple, nonradioactive, high throughput-compatible format for assaying the enzyme activity of glycosyltransferases that use diphosphonucleotide sugars as donor substrates
Novus Biologicals™ Maleimide Activated R-Phycoerythrin
For conjugation of RPhycoerythrin to proteins, peptides and ligands that contain sulfhydryl (-SH) groups
Novus Biologicals™ Maleimide Activated BSA
For conjugation of BSA to proteins, peptides and ligands that contain sulfhydryl (-SH) groups
Novus Biologicals™ PiBind™ Resin
PiBind™ resin provides quick and easy way to remove contaminating Pi from buffers
EMD Millipore™ Novagen™ Cleavage Control Protein Available on GSA/VA Contract for Federal Government customers only.
Monitors performance of thrombin or enterokinase cleavage conditions.
EMD Millipore™ Novagen™ Ekapture™ Agarose Available on GSA/VA Contract for Federal Government customers only.
Intended for rapid affinity-based capture.
Promega™ PinPoint™ Xa Protein Purification System
Designed for the production and purification of fusion proteins that are biotinylated in vivo. The DNA coding for the protein of interest is cloned into a PinPoint™ Vector downstream of a sequence encoding a peptide that becomes biotinylated in vivo.
Promega PinPoint Xa-1 Vector, 10microg
The PinPoint Xa Protein Purification System is designed for the production and purification of biotinylated fusion proteins.
Promega™ HaloTag™ Mammalian Pull-Down Systems
To capture and purify intracellular binary and higher order protein complexes, including transient or weakly interacting partners
Expedeon Amintra Heparin Resin
An affinity chromatography medium that is used for the purification of biomolecules that show affinity to heparin, such as antithrombin III, coagulation factors and other plasma proteins
  spinner