Quantitative Protein Interaction Reagents and Kits
Invitrogen™ Molecular Probes™ NanoOrange™ Protein Quantitation Kit, 200-2,000 assays Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Sensitive and easy assay for protein quantitation, with detection as low as 10 ng/mL of protein in solution
Invitrogen™ Molecular Probes™ CBQCA Protein Quantitation Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Extremely sensitive assay for quantitating proteins in solution, capable of detection as low as 10ng of protein per mL
Invitrogen™ Molecular Probes™ EZQ™ Protein Quantitation Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Fluorescence-based protein assay facilitates fast quantitation of protein samples prepared for gel electrophoresis
Invitrogen™ Molecular Probes™ ATTO-TAG™ CBQCA Derivatization Reagent (CBQCA; 3-(4-Carboxybenzoyl)quinoline-2-Carboxaldehyde) Non-distribution product as customer accommodation.
Reacts specifically with primary amines to form conjugates that can be analyzed by electrophoretic or chromatographic methods
Promega™ NanoBRET™ Nano-Glo™ Detection System
Dedicated detection for NanoBRET™ protein interaction assays.
Invitrogen™ Molecular Probes™ ATTO-TAG™ FQ Amine-Derivatization Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Reacts specifically with primary amines to form conjugates that can be analyzed by electrophoretic or chromatographic methods
Promega™ NanoBRET™ Nano-Glo™ Substrate
Replacement reagent to use with NanoBRET™ system.
Thermo Scientific™ Pierce™ qIP Protein Interaction Assay Reagents
Prepare optimized buffers for pull-down and reagent sets of TurboLuc (Tluc) assay buffer and coelenterazine substrate for luciferase quantitation with these reagents.
Thermo Scientific™ Expression Vectors for Pierce™ qIP Kits
Optimize cloning with this group of vectors, which includes epitope-tagged (HA and c-Myc) and TurboLuc-tagged (Tluc) multiple cloning site mammalian expression vectors.
Thermo Scientific™ Pierce™ Magnetic qIP Protein Interaction Kit, Tluc and Myc Tags
Pull down (co-IP) and measure interactions between c-Myc-tagged and Tluc-tagged protein pairs expressed in mammalian cells.
Thermo Scientific™ Pierce™ Magnetic qIP Protein Interaction Kit, Tluc and HA Tags
Pull down (co-IP) and measure interactions between HA-tagged and Tluc-tagged protein pairs expressed in mammalian cells with this kit.
Thermo Scientific™ Pierce™ Agarose qIP Protein Interaction Kit, Tluc and Myc Tags
Pull down (co-IP) and measure interactions between c-Myc-tagged and Tluc-tagged protein pairs expressed in mammalian cells. Thermo Scientific™ Pierce™ Agarose qIP Protein Interaction Kit, Tluc and Myc Tags uses anti-c-Myc agarose beads and a sensitive luciferase assay system.
Thermo Scientific™ Pierce™ Agarose qIP Protein Interaction Kit, Tluc and HA Tags
Pull down (co-IP) and measure interactions between HA-tagged and Tluc-tagged protein pairs expressed in mammalian cells with this kit.
Promega™ NanoBRET™ KRas/BRaf Interaction Assay
Sensitive live cell assays for screening cell signaling protein interactions.
Promega™ NanoBRET™ BRD4/Histone H4 Interaction Assay
Protein interaction assays to study Bromodomain/Histone interactions in live cells. The NanoBRET™ assays enable interaction studies in the context of natural chromatin. In addition to the full-length proteins, the BRD4 fragment is also included to study the interaction of this isolated domain.
Promega™ NanoBRET™ BRD9/Histone H4 Interaction Assay
Protein interaction assays to study Bromodomain/Histone interactions in live cells. The NanoBRET™ assays enable interaction studies in the context of natural chromatin. In addition to the full-length proteins, the BRD9 fragment is also included to study the interaction of this isolated domain.
Promega™ NanoBRET™ BRPF1/Histone H3.3 Interaction Assay
Protein interaction assays to study Bromodomain/Histone interactions in live cells. The NanoBRET™ assays enable interaction studies in the context of natural chromatin. In addition to the full-length proteins, the BRPF1 fragment is also included to study the interaction of this isolated domain.
Promega™ NanoBRET™ BRD4/Histone H3.3 Interaction Assay
Protein interaction assays to study Bromodomain/Histone interactions in live cells. The NanoBRET™ assays enable interaction studies in the context of natural chromatin. In addition to the full-length proteins, the BRD4 fragment is also included to study the interaction of this isolated domain.
Promega™ NanoBRET™ PPI Control Pair (p53, MDM2)
The NanoBRET™ PPI Control Pair (p53, MDM2) is an optimized pair of vectors designed to measure the interaction of protein partners p53 and MDM2 using the NanoBRET™ assay.
Promega™ NanoBRET™ BRPF1/Histone H4 Interaction Assay
Protein interaction assays to study Bromodomain/Histone interactions in live cells. The NanoBRET™ assays enable interaction studies in the context of natural chromatin. In addition to the full-length proteins, the BRPF1 fragment is also included to study the interaction of this isolated domain.
Promega™ NanoBRET™ Positive Control
The NanoBRET™ Positive Control Vector encodes a NanoLuc™-HaloTag™ fusion protein that tethers together the NanoLuc™ donor and HaloTag™ acceptor proteins to ensure efficient energy transfer.
Promega™ NanoBRET™ cMyc/MAX Interaction Assay
Sensitive live-cell assays for monitoring the interaction of proteins involved in transcription regulation.
Promega™ NanoBRET™ BRD9/Histone H3.3 Interaction Assay
Protein interaction assays to study Bromodomain/Histone interactions in live cells. The NanoBRET™ assays enable interaction studies in the context of natural chromatin. In addition to the full-length proteins, the BRD9 fragment is also included to study the interaction of this isolated domain.
Promega™ NanoBRET™ PPI Flexi™ Starter System
BRET-based assays to detect Protein:Protein interactions in live cells. The NanoBRET™ PPI Starter Systems utilize reversible assay technology. Assays can be performed in 96- or 384-well formats with low variability and high reproducibility.
Promega™ NanoBRET™ PPI MCS Starter System
BRET-based assays to detect Protein:Protein interactions in live cells. The NanoBRET™ PPI Starter Systems utilize reversible assay technology. Assays can be performed in 96- or 384-well formats with low variability and high reproducibility.
  spinner