Tag-Based IP and Pull-Down Kits
Thermo Scientific™ Pierce™ HA Tag IP/Co-IP Kit Available on GSA/VA Contract for Federal Government customers only.
Anti-HA antibody agarose beads and complete reagent set to easily co-immunoprecipitate protein interactions involving HA-tagged fusion proteins.
Thermo Scientific™ Pierce™ GST Protein Interaction Pull-Down Kit
Easily capture and affinity purify binding partners of GST-tagged fusion proteins with these glutathione (GSH) agarose beads and complete reagent set.
Thermo Scientific™ Pierce™ PolyHis™ Protein Interaction Pull-Down Kit Available on GSA/VA Contract for Federal Government customers only.
Easily capture and affinity purify binding partners of His-tagged fusion proteins using nickel-chelate (Ni-IDA) agarose beads and this complete reagent set.
Thermo Scientific™ Pierce™ c-Myc Tag IP/Co-IP Kit
Anti-c-Myc antibody agarose beads and complete reagent set to easily co-immunoprecipitate protein interactions involving c-Myc-tagged fusion proteins.
Thermo Scientific™ Pierce™ Biotinylated Protein Interaction Pull-Down Kit Available on GSA/VA Contract for Federal Government customers only.
Easily capture and affinity-purify binding partners of any biotin-labeled protein or other ligand using these streptavidin agarose beads and complete reagent set.
Thermo Scientific™ Pierce™ HA-Tag Magnetic IP/Co-IP Kit Available on GSA/VA Contract for Federal Government customers only.
Perform immunoprecipitation (IP) assays of HA-tagged proteins or co-IP reactions with HA-tagged bait proteins with these magnetic beads, positive control and reagents.
Thermo Scientific™ Pierce™ c-Myc-Tag Magnetic IP/Co-IP Kit
Magnetic beads for efficient isolation of c-Myc-tagged proteins and interactors.
Thermo Scientific™ Pierce™ c-Myc Tag IP/Co-IP Kit
Uses high-affinity antibodies to efficiently isolate c-Myc-tagged proteins and interactors
Thermo Scientific™ EZ-Link™ Desthiobiotinylation and Pull-Down Kit
Label purified proteins for bait-and-capture assays using streptavidin beads.
Promega™ HaloTag™ Complete Pull-Down System
Identify more physiologically relevant protein partners. HaloTag® Complete Pull-Down System includes buffers, resin, the TMRDirect™ Ligand, a starter cloning system, Wizard™ SV Gel and PCR Clean-Up System, and FuGENE™ HD Transfection Reagent.
  spinner