Protein Resins and Kits
Thermo Scientific™ Pierce™ Protein L Agarose and Kits
Affinity purify antibodies that have kappa light chains using Protein L offered in bottles, spin columns, filter plates and complete purification kits.
Thermo Scientific™ Pierce™ High-Capacity Protein A/G Agarose Available on GSA/VA Contract for Federal Government customers only.
Purify antibodies or perform immunoprecipitation with this exceptionally high-capacity, recombinant Protein A/G beaded agarose.
Thermo Scientific™ Pierce™ Protein A/G Agarose and Kits Available on GSA/VA Contract for Federal Government customers only.
Affinity purify antibodies (IgG) with versatile, recombinant Protein A/G agarose offered in bottles, spin columns, filter plates and complete purification kits.
Thermo Scientific™ Pierce™ Protein G Agarose and Kits Available on GSA/VA Contract for Federal Government customers only.
Affinity purify antibodies (IgG) with immobilized recombinant Protein G agarose offered in bottles, spin columns, filter plates and complete purification kits.
Thermo Scientific™ Pierce™ High-Capacity Protein A Agarose and Kits Available on GSA/VA Contract for Federal Government customers only.
Affinity purify antibodies (IgG) with densely immobilized Protein A offered in bottles, spin columns, filter plates and complete purification kits.
Thermo Scientific™ Pierce™ High-Capacity Protein L Agarose
Purify antibodies containing kappa light chains or perform immunoprecipitation with this high-capacity Protein L resin on beaded agarose.
Thermo Scientific™ Pierce™ High-Capacity Protein A/G UltraLink™ Resin
Extremely durable polyacrylamide support with densely immobilized, recombinant Protein A/G for exceptional IgG purification.
GE Healthcare HiTrap™ Protein G HP Prepacked Columns Available on GSA/VA Contract for Federal Government customers only.
Use for fast, easy separations of IgG from different species and subclasses. GE Healthcare Life Sciences™ HiTrap™ Protein G HP Prepacked Columns are prepacked with Protein G Sepharose™.
GE Healthcare HiTrap™ Protein A HP Prepacked Columns Available on GSA/VA Contract for Federal Government customers only.
Use for purification and isolation of monoclonal and polyclonal IgG from ascites, serum and cell culture supernatants. GE Healthcare Life Sciences™ HiTrap™ Protein A HP Prepacked Columns are specially designed, together with the medium to provide fast, simple and easy separations in a convenient format.
Thermo Scientific™ Pierce™ Protein A Agarose Available on GSA/VA Contract for Federal Government customers only.
Purify antibodies or perform immunoprecipitation with this standard-capacity, immobilized native Protein A on beaded agarose resin.
GE Healthcare Protein A/G HP SpinTrap™ Buffer Kit Available on GSA/VA Contract for Federal Government customers only.
Pre-made buffers optimized for the affinity-based enrichment of a target protein/immunoprecipitation using Protein A HP, Protein G HP, and Ab SpinTrap™ columns
Thermo Scientific™ Pierce™ Protein A/G UltraLink™ Resin Available on GSA/VA Contract for Federal Government customers only.
Extremely durable polyacrylamide support with immobilized recombinant Protein A/G for various IgG purification methods.
Thermo Scientific™ Pierce™ Recombinant Protein A Agarose Resin Available on GSA/VA Contract for Federal Government customers only.
Purify IgG using this genetically engineered, immobilized protein A on beaded agarose, when native protein A has undesirable effects.
GE Healthcare GammaBind™ G Sepharose™ Medium Available on GSA/VA Contract for Federal Government customers only.
Use recombinant GammaBind™ for laboratory scale antibody purification. GE Healthcare Life Sciences™ GammaBind™ G Sepharose™ Medium binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse
Promega Magne™ Protein A Beads, 20% Slurry
For purification of immunoglobulins from cell culture media, ascites and serum samples. These beads offer a convenient method for achieving high purity and high recovery of monoclonal and polyclonal antibodies from a variety of biological samples.
MilliporeSigma™ Upstate Protein G Agarose, Fast Flow
Used for medium low pressure chromatography of IgG from mouse, sheep, rabbit, for immunoprecipitations.
GE Healthcare Protein A HP MultiTrap™ Plates
For small-scale purification of antibodies or enrichment of proteins of interest by antibody capture in 96-well filter plates
MilliporeSigma™ Upstate Protein A Agarose
Used for purification of human, mouse rabbit immunoglobins.
GE Healthcare Protein A HP SpinTrap™ Column
For simple, small-scale antibody purification or enrichment of specific proteins by antibody capture in single-use spin columns
Thermo Scientific™ Pierce™ High-Capacity Protein G Agarose Available on GSA/VA Contract for Federal Government customers only.
High-capacity, recombinant Protein G beaded agarose resin for a variety of antibody affinity purification methods.
MilliporeSigma™ Upstate Protein G Agarose/Salmon Sperm DNA
Blocks non-specific DNA binding sites on agarose beads.
MilliporeSigma™ Calbiochem™ PANSORBIN™ Cells, Standardized Available on GSA/VA Contract for Federal Government customers only.
For mitogenic stimulation of B lyphocytes and for immunoprecipitation
Thermo Scientific™ Pierce™ High-Capacity Protein A UltraLink™ Resin
Extremely durable polyacrylamide support with densely immobilized Protein A for high-capacity IgG purification.
SouthernBiotech™ Protein G-Unlabeled
SouthernBiotech binding proteins are useful for immunoprecipitation, purification, and ELISA applications.
Novex™ Recombinant Protein G - Sepharose™ 4B Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Recombinant Protein G - Sepharose™ 4B
GE Healthcare HiTrap™ rProtein A FF Columns Available on GSA/VA Contract for Federal Government customers only.
Use for preparative purification of monoclonal and polyclonal antibodies from cell culture supernatants, serum and ascites. GE Healthcare Life Sciences™ HiTrap™ rProtein A FF Columns are prepacked with rProtein A Sepharose™ Fast Flow.
Novex™ Recombinant Protein A-Sepharose™ 4B Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Recombinant Protein A-Sepharose™ 4B
Thermo Scientific™ Pierce™ Protein G UltraLink™ Resin
Extremely durable polyacrylamide support with immobilized, recombinant Protein G for various IgG purification methods.
MilliporeSigma™ Upstate Protein A Agarose/Salmon Sperm DNA Available on GSA/VA Contract for Federal Government customers only.
Blocks non-specific DNA binding sites on agarose beads.
Novex™ Protein A - Sepharose™ 4B Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Protein A - Sepharose™ 4B
  spinner