Protein A Resins and Kits
Thermo Scientific™ Pierce™ Protein A Agarose Available on GSA/VA Contract for Federal Government customers only.
Purify antibodies or perform immunoprecipitation with this standard-capacity, immobilized native Protein A on beaded agarose resin.
GE Healthcare HiTrap™ Protein A HP Prepacked Columns Available on GSA/VA Contract for Federal Government customers only.
Use for purification and isolation of monoclonal and polyclonal IgG from ascites, serum and cell culture supernatants. GE Healthcare Life Sciences™ HiTrap™ Protein A HP Prepacked Columns are specially designed, together with the medium to provide fast, simple and easy separations in a convenient format.
Thermo Scientific™ Pierce™ High-Capacity Protein A Agarose and Kits Available on GSA/VA Contract for Federal Government customers only.
Affinity purify antibodies (IgG) with densely immobilized Protein A offered in bottles, spin columns, filter plates and complete purification kits.
Novex™ Protein A - Sepharose™ 4B Available on GSA/VA Contract for Federal Government customers only.
Protein A - Sepharose™ 4B
Thermo Scientific™ Pierce™ High-Capacity Protein A UltraLink™ Resin
Extremely durable polyacrylamide support with densely immobilized Protein A for high-capacity IgG purification.
GE Healthcare Protein A HP SpinTrap™ Column Available on GSA/VA Contract for Federal Government customers only.
For simple, small-scale antibody purification or enrichment of specific proteins by antibody capture in single-use spin columns
EMD Millipore™ Upstate Protein A Agarose/Salmon Sperm DNA Available on GSA/VA Contract for Federal Government customers only.
Blocks non-specific DNA binding sites on agarose beads.
Thermo Scientific™ Pierce™ Recombinant Protein A Agarose Resin Available on GSA/VA Contract for Federal Government customers only.
Purify IgG using this genetically engineered, immobilized protein A on beaded agarose, when native protein A has undesirable effects.
Novex™ Recombinant Protein A-Sepharose™ 4B Available on GSA/VA Contract for Federal Government customers only.
Recombinant Protein A-Sepharose™ 4B
Thermo Scientific™ Pierce™ Protein A UltraLink™ Resin
Extremely durable polyacrylamide support with immobilized Protein A for standard-capacity IgG purification.
Alfa Aesar™ Antibody Purification on Protein A Column
Buffers to be used when performing antibody purification using immobilized Protein A
GE Healthcare HRP labelled Protein A Available on GSA/VA Contract for Federal Government customers only.
Protein A conjugated with Horseradish peroxidase for detection of immunoglobulins via affinity to the Fc region - typically IgG from many mammalian species
MP Biomedicals™ Protein A, Fc Binding Grade
Fc Binding Grade, White Lyophilized Powder. Routinely used for immunoprecipitation assays.
GE Healthcare HiTrap™ rProtein A FF Columns Available on GSA/VA Contract for Federal Government customers only.
Use for preparative purification of monoclonal and polyclonal antibodies from cell culture supernatants, serum and ascites. GE Healthcare Life Sciences™ HiTrap™ rProtein A FF Columns are prepacked with rProtein A Sepharose™ Fast Flow.
MP Biomedicals™ Protein A-HRP
Use this versatile reagent in many immunological assay systems
MP Biomedicals™ Protein A, Recombinant, Purity: 98%
Useful for the study of cell surface antigens and receptors, and for detecting antibody-secreting cells
GE Healthcare Protein A HP MultiTrap™ Plates Available on GSA/VA Contract for Federal Government customers only.
For small-scale purification of antibodies or enrichment of proteins of interest by antibody capture in 96-well filter plates
EMD Millipore™ Upstate Protein A Agarose Available on GSA/VA Contract for Federal Government customers only.
Used for purification of human, mouse rabbit immunoglobins.
EMD Millipore™ Calbiochem™ Protein A Agarose Suspension Available on GSA/VA Contract for Federal Government customers only.
Used in immunoprecipitation applications
Abnova™ TCS Antibody Purification Kit
For measuring Trypsin activity using fluorometric methods
Abnova™ Serum Antibody Purification Kit
For measuring Trypsin activity using fluorometric methods
Abnova™ Antibody Purification Kit
For measuring Trypsin activity using fluorometric methods
Expedeon Amintra Protein A Magnetic Resin
For batch magnetic purification of immunoglobulins
Novex™ Rec Protein A-HRP Available on GSA/VA Contract for Federal Government customers only.
Rec Protein A-HRP
  spinner