Protein G Resins and Kits
Thermo Scientific™ Pierce™ High-Capacity Protein G Agarose Available on GSA/VA Contract for Federal Government customers only.
High-capacity, recombinant Protein G beaded agarose resin for a variety of antibody affinity purification methods.
Thermo Scientific™ Pierce™ Protein G Agarose and Kits Available on GSA/VA Contract for Federal Government customers only.
Affinity purify antibodies (IgG) with immobilized recombinant Protein G agarose offered in bottles, spin columns, filter plates and complete purification kits.
Thermo Scientific™ Pierce™ Protein G UltraLink™ Resin
Extremely durable polyacrylamide support with immobilized, recombinant Protein G for various IgG purification methods.
GE Healthcare HiTrap™ Protein G HP Prepacked Columns Available on GSA/VA Contract for Federal Government customers only.
Use for fast, easy separations of IgG from different species and subclasses. GE Healthcare Life Sciences™ HiTrap™ Protein G HP Prepacked Columns are prepacked with Protein G Sepharose™.
GE Healthcare GammaBind™ G Sepharose™ Medium Available on GSA/VA Contract for Federal Government customers only.
Use recombinant GammaBind™ for laboratory scale antibody purification. GE Healthcare Life Sciences™ GammaBind™ G Sepharose™ Medium binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse
Novex™ Recombinant Protein G - Sepharose™ 4B Available on GSA/VA Contract for Federal Government customers only.
Recombinant Protein G - Sepharose™ 4B
EMD Millipore™ Upstate Protein G Agarose, Fast Flow Available on GSA/VA Contract for Federal Government customers only.
Used for medium low pressure chromatography of IgG from mouse, sheep, rabbit, for immunoprecipitations.
Promega™ Magne™ Protein A Beads, 20% Slurry
For purification of immunoglobulins from cell culture media, ascites and serum samples. These beads offer a convenient method for achieving high purity and high recovery of monoclonal and polyclonal antibodies from a variety of biological samples.
EMD Millipore™ Upstate Protein G Agarose/Salmon Sperm DNA Available on GSA/VA Contract for Federal Government customers only.
Blocks non-specific DNA binding sites on agarose beads.
Thermo Scientific™ Pierce™ High-Capacity Protein G UltraLink™ Resin
Extremely durable polyacrylamide support with densely immobilized, recombinant Protein G for various IgG purification methods.
Novex™ Recombinant Protein G-HRP Available on GSA/VA Contract for Federal Government customers only.
Recombinant Protein G-HRP
Novex™ Recombinant Protein G Available on GSA/VA Contract for Federal Government customers only.
Recombinant Protein G
Thermo Scientific™ POROS™ MabCapture™ A/G Select
MabCapture A/G Select allows antibody purification with high dynamic binding performance at low linear flow rates and maintains superior binding capacity at high linear flow rates
Thermo Scientific™ POROS™ MabCapture™ A/G Select
MabCapture A/G Select allows antibody purification with high dynamic binding performance at low linear flow rates and maintains superior binding capacity at high linear flow rates
Thermo Scientific™ POROS™ MabCapture™ A/G Select
MabCapture A/G Select allows antibody purification with high dynamic binding performance at low linear flow rates and maintains superior binding capacity at high linear flow rates
Thermo Scientific™ Pierce™ Protein A/G Plus Agarose
Thermo Scientific Pierce Protein A/G Plus Agarose is an exceptionally high-capacity Protein A/G beaded agarose resin for use in a variety of antibody affinity purification methods
MP Biomedicals™ Protein G
Recombinant Protein
GE Healthcare HiTrap™ Protein G HP Prepacked Columns Promotion
For a limited time only, receive a 10% discount on your next HiTrap order
  spinner