Protein L Resins and Kits
Thermo Scientific™ Pierce™ Protein L Agarose and Kits
Affinity purify antibodies that have kappa light chains using Protein L offered in bottles, spin columns, filter plates and complete purification kits.
Thermo Scientific™ Pierce™ High-Capacity Protein L Agarose
Purify antibodies containing kappa light chains or perform immunoprecipitation with this high-capacity Protein L resin on beaded agarose.
  spinner