Ultrafiltration and Centrifugal Devices
EMD Millipore™ BioPak™ Disposable Ultrafilter Available on GSA/VA Contract for Federal Government customers only.
Nuclease-free, pyrogen-free water from any EMD Millipore Ultrapure™ water system
EMD Millipore™ Pellicon™ XL Devices Available on GSA/VA Contract for Federal Government customers only.
Small-volume tangential flow filter device
EMD Millipore™ Ultracel™ Ultrafiltration Disc Available on GSA/VA Contract for Federal Government customers only.
Use to concentrate or desalt dilute solutions with the Stirred Cell device
Thermo Scientific™ Pierce™ PES Protein Concentrators Available on GSA/VA Contract for Federal Government customers only.
Concentrate and desalt biological samples and perform buffer exchange with disposable polyethersulfone (PES)-membrane ultrafiltration centrifugal devices.
Sartorius™ Vivaflow 200 Crossflow Casettes Available on GSA/VA Contract for Federal Government customers only.
Offers a standard for ease of use, reliability and flexibility unrivalled by other filtration systems. Sartorius™ Vivaflow 200 Crossflow Cassettes are ready to use, reusable laboratory crossflow cassettes for concentrating up to 5L samples. Available in different MWCOs and membrane types for different application needs.
Sartorius™ Vivaflow™ 50 Crossflow Casettes Available on GSA/VA Contract for Federal Government customers only.
Single use. Sartorius™ Vivaflow™ 50 Crossflow Casettes are ready-to-use for filtration and concentration of samples from 100 mL to 5L.
Corning™ Spin-X™ UF Concentrators Available on GSA/VA Contract for Federal Government customers only.
Disposable, single use only ultrafiltration devices with polyethersulfone membranes (PES) for the centrifugal concentration and/or purification of biological fluids and aqueous solutions
Sartorius™ Vivacon™ 2 DNA Concentrator Available on GSA/VA Contract for Federal Government customers only.
Centrifugal units for fast DNA concentration and buffer exchange of 2 ml samples
EMD Millipore™ Millex-GV Filter for Samplicity G2, 0.22μm, PVDF, 33mm, Nonsterile
Used for non-sterile filtration on the Samplicity™ G2 system. This 33mm syringe filter with a hydrophilic Durapore™ PVDF membrane and 0.22μm pore size is used to prepare solutions prior to chromatography or other instrument analyses.
EMD Millipore™ Ultrafiltration Membranes - BioMax™ PB21 Available on GSA/VA Contract for Federal Government customers only.
High-recovery membranes for stirred cells
EMD Millipore™ Millex-HV Filter for Samplicity G2, 0.45μm, PVDF, 33mm, Nonsterile
Used for non-sterile filtration on the Samplicity™ G2 system. This 33mm syringe filter with a hydrophilic Durapore™ PVDF membrane and 0.45μm pore size is used to prepare solutions prior to chromatography or other instrument analyses.
EMD Millipore™ Biomax™ Ultrafiltration Membrane Discs Available on GSA/VA Contract for Federal Government customers only.
Recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media. EMD Millipore Biomax Ultrafiltration Membrane Discs are compatible with Amicon™ Stirred Cells.
Sartorius™ Vivaflow™ 50R Crossflow Casettes
Unique, Plug and Play laboratory crossflow devices for concentrating samples like viruses and antibodies
Sartorius™ Collection Tubes for Vivacon™ 500, Bulk Pack (100 pcs)
Bulk pack (100 pieces) of collection reservoirs for use with Vivacon 500 centrifugal units.
EMD Millipore™ Prep/Scale Spiral-Wound Ultrafiltration Modules: TFF-2 Available on GSA/VA Contract for Federal Government customers only.
For easy, reliable preparation of samples ranging from 100L down to 100mL
EMD Millipore™ Samplicity™ G2 Filtration System Replacement Lid
To be used with the Samplicity G2 Filtration System.
EMD Millipore™ Samplicity™ G2 Filtration System
Vacuum filtration system using Millex™ filters for sample preparation for HPLC.
EMD Millipore™ Millex Samplicity™ Filters, 0.45μm Hydrophilic PVDF Available on GSA/VA Contract for Federal Government customers only.
To be used with the Samplicity Filtration System. Powered by vacuum, the system takes minimal manual effort to filter 1 to 8 samples into standard HPLC vials in seconds, with higher recovery than syringe filters provide.
EMD Millipore™ Millex-GN Filters for Samplicity G2, 0.20um, Nylon, 33mm, Nonsterile
Used for non-sterile filtration on the Samplicity™ G2 system. This 33mm syringe filter with a hydrophilic nylon membrane and 0.2μm pore size is used to prepare solutions prior to chromatography or other instrument analyses.
  spinner