Total Protein Assay Kits
Thermo Scientific™ Pierce™ BCA™ Protein Assay Available on GSA/VA Contract for Federal Government customers only.
Green-to-blue (A562nm), precise, detergent-compatible assay reagent to measure total protein concentration vs. protein standard.
Thermo Scientific™ Pierce™ Bovine Serum Albumin Standards Available on GSA/VA Contract for Federal Government customers only.
High-quality, calibrated BSA solutions (2mg/mL) to serially dilute to create protein assay standard curves to accurately measure protein concentration.
Thermo Scientific™ Pierce™ Micro BCA™ Protein Assay Available on GSA/VA Contract for Federal Government customers only.
Special 3-component version of our popular BCA Reagents to measure total protein concentration of dilute protein solutions (0.5 to 20 micrograms/mL).
Thermo Scientific™ Pierce™ Coomassie Plus™ (Bradford) Protein Assay Available on GSA/VA Contract for Federal Government customers only.
Brown-to-purple (A595nm), ready-to-use, fast reducing agent-compatible assay reagent to measure total protein concentration vs. protein standard.
Thermo Scientific™ Pierce™ Coomassie (Bradford) Protein Assay Available on GSA/VA Contract for Federal Government customers only.
Brown-to-purple (A595nm), ready-to-use, classic reducing agent-compatible assay reagent to measure total protein concentration vs. protein standard.
Thermo Scientific™ Pierce™ 660nm Protein Assay Available on GSA/VA Contract for Federal Government customers only.
Red-to-green (A660nm), ready-to-use, detergent- and reducing agent-compatible assay reagent to measure total protein concentration vs. protein standard.
Thermo Scientific™ Pierce™ Modified Lowry Protein Assay
Clear-to-purple (A750nm), 2-component, folin phenol- and copper-based reagent system to measure total protein concentration vs. protein standard.
Thermo Scientific Pierce™ Rapid Gold BCA Protein Assay Kit
Provides high accuracy results in fraction of time. Thermo Scientific™ Pierce™ Rapid Gold BCA Protein Assay Kit is two-component, high-precision, detergent-compatible assay optimized to measure (at 480nm) total protein concentration compared to protein standard curve of known concentrations.
Thermo Scientific™ Pierce™ BCA™ Protein Assay, Reducing Agent Compatible (RAC) Available on GSA/VA Contract for Federal Government customers only.
Reducing agent-compatible version of our popular BCA Reagent Kit to measure protein concentration in samples containing DTT or BME thiol-reductants.
Thermo Scientific™ Pierce™ Detergent Compatible Bradford Assay Available on GSA/VA Contract for Federal Government customers only.
Quickly and reliably measure protein concentration of cell lysates or other samples that contain Triton™ X-100, NP-40 or other detergents using the the Thermo Scientific™ Pierce™ Detergent Compatible Bradford Assay Kit.
Thermo Scientific™ Pierce™ Bovine Gamma Globulin Standards
High-quality, calibrated BGG solutions (2mg/mL) to serially dilute to create protein assay standard curves to measure antibody (IgG) concentration.
Thermo Scientific™ Pierce™ Compat-Able™ Protein Assay Kits
Precipitation reagents to eliminate interfering substances from protein assay samples to make them compatible with BCA and Coomassie-Bradford assays.
Alfa Aesar™ BCA Protein Assay Kit
Contains two separate solutions for estimation of protein concentration
Thermo Scientific™ Pierce™ Detergent Compatible Bradford Assay Reagent
The Thermo Scientific™ Pierce™ Detergent Compatible Bradford Assay Reagent is a quick and ready-to-use modification of the well-known Bradford coomassie dye-binding, colorimetric method for total protein quantitation
EMD Millipore™ Novagen™ BCA Protein Assay Kit Available on GSA/VA Contract for Federal Government customers only.
Determines protein concentration in standard assay or microassay configuration.
Thermo Fisher Scientific™ Invitrogen™ ProQuantum™ Human IL-6 Immunoassay Kit
Measure cytokine proteins with small sample consumption and high sensitvity on a qPCR instrument
Thermo Fisher Scientific™ Invitrogen™ ProQuantum™ Human IL-8 Immunoassay Kit
Measure cytokine proteins with small sample consumption and high sensitvity on a qPCR instrument
Abnova™ Total Histone H3 Acetylation Detection Fast Kit (Fluorometric)
Total Histone H3 Acetylation Detection Fast Kit (Fluorometric) is used for measuring total acetylation of histone H3.
Abnova™ Total Histone H4 Acetylation Detection Fast Kit (Colorimetric)
Total Histone H4 Acetylation Detection Fast Kit (Colorimetric) is used for measuring total acetylation of histone H4.
Abnova™ Total Histone H4 Acetylation Detection Fast Kit (Fluorometric)
Total Histone H4 Acetylation Detection Fast Kit (Fluorometric) is used for measuring total acetylation of histone H4.
Abnova™ Total Histone H3 Acetylation Detection Fast Kit (Colorimetric)
Total Histone H3 Acetylation Detection Fast Kit (Colorimetric) is used for measuring total acetylation of histone H3.
Abnova™ Histone H3 (Total) Quantification Kit (Fluorometric) Available on GSA/VA Contract for Federal Government customers only.
Histone H3 (Total) Quantification Kit (Fluorometric) is designed for quantifying levels of histone H3 proteins independent of its modified state and can also be used for normalizing the modified histone H3 content of samples when run in parallel with other histone modification quantification kit series.
Abnova™ Histone H3 (Total) Quantification Kit (Colorimetric) Available on GSA/VA Contract for Federal Government customers only.
Histone H3 (Total) Quantification Kit (Colorimetric) is designed for quantifying levels of histone H3 proteins independent of its modified state and can also be used for normalizing the modified histone H3 content of samples when run in parallel with other histone modification quantification kit series.
Thermo Fisher Scientific™ Invitrogen™ ProQuantum™ Human IFNγ Immunoassay Kit
Measure cytokine proteins with small sample consumption and high sensitvity on a qPCR instrument
Thermo Fisher Scientific™ Invitrogen™ ProQuantum™ Human IL-12p40 Immunoassay Kit
Measure cytokine proteins with small sample consumption and high sensitvity on a qPCR instrument
Thermo Fisher Scientific™ Invitrogen™ ProQuantum™ Human IL-1β Immunoassay Kit
Measure cytokine proteins with small sample consumption and high sensitvity on a qPCR instrument
Thermo Fisher Scientific™ Invitrogen™ ProQuantum™ Human IP-10 Immunoassay Kit
Measure cytokine proteins with small sample consumption and high sensitvity on a qPCR instrument
  spinner