Transcription Factor Assay Kits
Thermo Scientific™ TranscriptAid T7 High Yield Transcription Kit Available on GSA/VA Contract for Federal Government customers only.
Get high yield in vitro transcription from DNA templates containing a T7 RNA Polymerase promoter, including linearized plasmids or PCR products.
Abnova™ CREB1 (phospho S133) Transcription Factor Assay Kit
CREB1 (phospho S133) Transcription Factor Assay Kit is a colorimetric method for determining CREB1 DNA binding activity.
Abnova™ WST-1 Cell Proliferation Assay Kit Available on GSA/VA Contract for Federal Government customers only.
WST-1 Cell Proliferation Assay Kit provides an easy to use tool for studying induction and inhibition of cell proliferation in any in vitro model.
Abnova™ NF-kB (human p50) Transcription Factor Assay Kit
NF-kB (human p50) Transcription Factor Assay Kit is a non-radioactive method for detecting specific transcription factor DNA binding activity.
Abnova™ NF-kB (p65) Transcription Factor Assay Kit
NF-kB (p65) Transcription Factor Assay Kit is a non-radioactive method for detecting specific transcription factor DNA binding activity.
Abnova™ TP53 Transcription Factor Assay Kit
TP53 Transcription Factor Assay Kit is a method for detecting specific transcription factor DNA binding activity in nuclear extracts.
Abnova™ SREBF1 Transcription Factor Assay Kit
SREBF1 Transcription Factor Assay Kit is a method for detecting SREBF1 transcription factor DNA binding activity in nuclear extracts.
Abnova™ PPARA/PPARD/PPARG Complete Transcription Factor Assay Kit
PPARA/PPARD/PPARG Complete Transcription Factor Assay Kit is a non-radioactive, sensitive method for detecting PPAR-alpha, delta, gamma transcription factor DNA binding activity in nuclear extracts and whole cell lysates.
Abnova™ SREBF2 Transcription Factor Assay Kit
SREBF2 Transcription Factor Assay Kit is a non-radioactive, sensitive method for detecting specific transcription factor DNA binding activity in nuclear extracts and whole cell lysates.
Abnova™ PPARD Transcription Factor Assay Kit
PPARD Transcription Factor Assay Kit is a non-radioactive, sensitive method for detecting PPARD transcription factor DNA binding activity in nuclear extracts.
Abnova™ PPARA Transcription Factor Assay Kit
PPARA Transcription Factor Assay Kit is a non-radioactive, sensitive method for detecting PPARA transcription factor DNA binding activity in nuclear extracts.
Abnova™ HIF1A Transcription Factor Assay Kit
HIF1A Transcription Factor Assay Kit is a colorimetric method for determining HIF-1-alpha DNA binding activity.
Abnova™ PPARG Transcription Factor Assay Kit
PPARG Transcription Factor Assay Kit is a non-radioactive, sensitive method for detecting PPARG transcription factor DNA binding activity in nuclear extracts.
  spinner