Membranes for Hybridization and Transfer
Thermo Scientific™ PVDF Transfer Membrane, 0.2 µm, 26.5 cm x 3.75 m Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific Pierce PVDF Transfer Membranes are made of high-quality polyvinylidene difluoride and provide high binding capacity for proteins and nucleic acids for Western, Southern, and Northern blotting methods. This PVDF membrane has a 0.2-micron pore size and is ideal for colorimetric and chemiluminescent detection.
EMD Millipore™ Immobilon™-P PVDF Transfer Membranes Available on GSA/VA Contract for Federal Government customers only.
Optimized for binding proteins transferred from a variety of gel matrices. EMD Millipore Immobilon™-P PVDF Transfer Membranes have superior handling, staining, and resistance than nitrocellulose. For use with Western blots and protein sequencing. Available in sheets or rolls.
EMD Millipore™ Immobilon™ FL PVDF Transfer Membranes Available on GSA/VA Contract for Federal Government customers only.
Designed specifically for Western blots and protein sequencing. EMD Millipore Immobilon™-FL PVDF Transfer Membranes are plain surfaced with 0.45μm pores and have lower background fluorescence than other membranes. Available in 10cm and 20cm squares and a 3.75m roll.
EMD Millipore™ Immobilon™ PVDF Transfer Membranes: PSQ Available on GSA/VA Contract for Federal Government customers only.
For Western blots and protein sequencing
GE Healthcare Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls Available on GSA/VA Contract for Federal Government customers only.
Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. GE Healthcare Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
GE Healthcare Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Rolls Available on GSA/VA Contract for Federal Government customers only.
Use for the best results with fluorescence Western blotting detection applications. GE Healthcare Life Sciences™ Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
GE Healthcare Amersham™ Hybond™ -XL Membranes Available on GSA/VA Contract for Federal Government customers only.
Use for optimal signal-to-noise ratios when using radioactively labeled probes. GE Healthcare Life Sciences™ Amersham™ Hybond™ -XL Membranes are novel, charged nylon membranes that have a greater concentration and a more even distribution of positive charge than other charged nylon membranes.
GE Healthcare Amersham™ Hybond™ P 0.45μm PVDF Membrane Available on GSA/VA Contract for Federal Government customers only.
Use for chemiluminescent and fluorescent detection methods for proteins of > Mr 20 000. GE Healthcare Life Sciences™ Amersham™ Hybond™ P 0.45μm PVDF Membrane is a 0.45 μm pore size hydrophobic membrane, with high physical strength, highly suitable for stripping and reprobing.
GE Healthcare Amersham™ Hybond™ -N+ Membranes Available on GSA/VA Contract for Federal Government customers only.
Use for the highest sensitivity in DNA and RNA blotting. GE Healthcare Life Sciences™ Amersham™ Hybond™ -N+ Membrane is a positively charged nylon membranes for use with radioactive and nonradioactive chemiluminescence and chemifluorescence detection.
GE Healthcare Whatman™ Grade GB003 Blotting Paper Available on GSA/VA Contract for Federal Government customers only.
Ensures that no contamination will occur during the transfer steps. Whatman™ Grade GB003 Blotting Paper eliminates sheet to sheet variations with precisely cut membrane sizes.
GE Healthcare Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Rolls Available on GSA/VA Contract for Federal Government customers only.
Use for robust protein blotting. GE Healthcare Life Sciences™ Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
GE Healthcare Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets Available on GSA/VA Contract for Federal Government customers only.
Use for high sensitivity, resolution, and low background for all labeling and detection systems, including chemiluminescent, fluorescenct, colorimetric and radioactive. GE Healthcare Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
Thermo Scientific™ Owl™ Blotting Paper Available on GSA/VA Contract for Federal Government customers only.
Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with this high-quality blotting paper.
GE Healthcare Amersham™ Hybond™-N Membranes Available on GSA/VA Contract for Federal Government customers only.
Use for high sensitivity applications in DNA and RNA blotting. GE Healthcare Life Sciences™Amersham™ Hybond™-N Membranes is a strong supported membrane that is inherently hydrophilic and requires no pre-wetting.
GE Healthcare Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.45μm BA85 Protran Circles Available on GSA/VA Contract for Federal Government customers only.
100% pure nitrocellulose to ensure the highest binding capacity possible
EMD Millipore™ Immobilon™-P Blotting Sandwiches Available on GSA/VA Contract for Federal Government customers only.
Convenient, time-saving option for preparing multiple blots
GE Healthcare Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Sheets Available on GSA/VA Contract for Federal Government customers only.
Provides exceptional handling characteristics, allowing reprobing
EMD Millipore™ Immobilon™ Blotting Filter Paper Available on GSA/VA Contract for Federal Government customers only.
Chromatography-grade blotting filter paper precut to the most popular Western blotting sizes
GE Healthcare Whatman™ Nytran™ SuperCharge Membranes Available on GSA/VA Contract for Federal Government customers only.
Cast uniformly on both sides of a support matrix, providing excellent symmetry and allowing membranes to lie flat without curling
GE Healthcare Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets Available on GSA/VA Contract for Federal Government customers only.
Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. GE Healthcare Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
GVS Polyvinylidene Fluoride Hybridization and Transfer Membranes
Designed for Western transfers, amino acid analyses, and protein sequencing procedures
MP Biomedicals™ Biotrans(+) Nylon Membranes, 30cmx3m Roll, 0.45um
Used for alkaline, Western, and Improved Southern Blots. Reinforced nylon 66 membranes for use in DNA, RNA and protein transfers. The physical characteristics of BIOTRANS membranes make them especially useful as a transfer medium.
GE Healthcare Whatman™ Gel Blotting Paper Grade GB003 Available on GSA/VA Contract for Federal Government customers only.
A thick paper ideal for lysis/denaturation of colony or plaque lifts and Western blots
EMD Millipore™ Immobilon-NC Transfer Membrane Available on GSA/VA Contract for Federal Government customers only.
Maximum sensitivity with minimal background, suitable for both chemiluminescent and radioactive detection systems
EMD Millipore™ Immobilon-E Transfer Membrane
MilliporeSigma™ Immobilon™-E provides the a PVDF blotting membrane that wets in transfer buffer or water and eliminates the need for the alcohol pre-wet step
GE Healthcare Whatman™ Grade GB005 Blotting Paper Available on GSA/VA Contract for Federal Government customers only.
Recommended for applications where fewer layers of blotting paper must still ensure a high capacity. Whatman™ Grade GB005 Blotting Paper ensures the wicking capability and uniformity of capillary action that are important in obtaining clean and even transfers during blotting.
GE Healthcare Amersham™ Hybond™ PVDF Membranes: Sheets Available on GSA/VA Contract for Federal Government customers only.
Ideal for Western blotting applications where stripping and reprobing are necessary
  spinner