RNAi Screening Libraries
Invitrogen™ Silencer™ Human Kinase siRNA Library Non-distribution product as customer accommodation.
The Silencer™ Human Kinase siRNA Library contains 2,127 Invitrogen™ Silencer siRNAs targeting 709 human kinase genes for performing RNAi screens against the kinome.
Invitrogen™ Silencer™ Human DNA Damage Response siRNA Library Non-distribution product as customer accommodation.
The Silencer™ Human DNA Damage Response siRNA Library contains 1,725 Invitrogen™ Silencer siRNAs targeting 575 genes for performing RNAi screens against known DNA damage response pathways.
Invitrogen™ Silencer™ Human Phosphatase siRNA Library Non-distribution product as customer accommodation.
Contains 894 Invitrogen™ Silencer siRNAs targeting 298 human phosphatase genes and plated onto twelve 96-well plates. Thermo Scientific Silencer™ Human Phosphatase siRNA Library is designed using the most up-to-date genome databases for use in RNAi, epigenetics and non-coding RNA research applications.
Invitrogen™ LentiArray™ Human Nuclear Hormone Receptor Library, Glycerol Stock Non-distribution product as customer accommodation.
Targets 47 genes with up to 4 gRNA per gene target for a total of 188 gRNAs. Libraries are delivered as 50μL of glycerol stock per gRNA and are also available as ready to use packaged lentivirus.
  spinner