Microbiology Equipment
Thermo Scientific™ AnaeroPack™-Anaero Anaerobic Gas Generator Available on GSA/VA Contract for Federal Government customers only.
Mitsubishi™ AnaeroPack-Anaero is used for cultivation of anaerobic microorganisms without the addition of water or use of a catalyst.
Thermo Scientific™ AnaeroPack™-MicroAero Gas Generator Available on GSA/VA Contract for Federal Government customers only.
Mitsubishi™ AnaeroPack-MicroAero is used for cultivation of microaerophilic microorganisms
Thermo Scientific™ AnaeroPack™ 7.0L Rectangular Jar Available on GSA/VA Contract for Federal Government customers only.
Mitsubishi™ AnaeroPack 7.0L Rectangular Jar is an anaerobic jar with easy-close lid.
Thermo Scientific™ AnaeroPouch™-Anaero Anaerobic Gas Generator Available on GSA/VA Contract for Federal Government customers only.
Mitsubishi™ AnaeroPouch-Anaero is used for cultivation of anaerobic microorganisms without the addition of water or use of a catalyst.
Thermo Scientific™ Oxoid™ AnaeroGen™ 2.5L Sachet Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Oxoid AnaeroGen 2.5L Sachets are anaerobic gas generating sachets for use with Thermo Scientific™ Oxoid 2.5L jar, Part No. AG0025A (not included).
Thermo Scientific™ AnaeroPack™-CO2 Generator Available on GSA/VA Contract for Federal Government customers only.
Mitsubishi™ AnaeroPack-CO2 is used for cultivation of CO2 dependent microorganisms
Thermo Scientific™ AnaeroPack™ 2.5L Rectangular Jar Available on GSA/VA Contract for Federal Government customers only.
Mitsubishi™ AnaeroPack 2.5L Rectangular Jar is an anaerobic jar with easy-close lid.
BD GasPak™ EZ Gas Generating Systems and Supplies: Pouch Systems Available on GSA/VA Contract for Federal Government customers only.
Used in producing anaerobic, microaerophilic, or CO2-enriched environments.   BD Difco™ GasPak™ EZ Gas Generating Systems and Supplies provide catalyst-free convenience.
BD GasPak™ EZ Gas Generating Systems and Supplies: Container System Sachets Available on GSA/VA Contract for Federal Government customers only.
Used in producing anaerobic, microaerophilic, or CO2-enriched environments.   BD Difco™ GasPak™ EZ Gas Generating Systems and Supplies: Container System Sachets provide catalyst-free convenience.
Thermo Scientific™ RT Anaero-Indicator Available on GSA/VA Contract for Federal Government customers only.
Mitsubishi™ RT Anaero-Indicator is a single-use disposable means of detecting an anaerobic atmosphere.
Eppendorf™ Femtotips™ Microloader Tips for Femtojet Microinjector Available on GSA/VA Contract for Federal Government customers only.
Tips to fill microinjection capillaries and recovering surplus solution from capillary
Eppendorf™ Femtotips™ Microinjection Capillary Tips Available on GSA/VA Contract for Federal Government customers only.
Disposable glass capillaries for reproducible microinjection into adherent and suspension cells
BD GasPak™ Supplies Available on GSA/VA Contract for Federal Government customers only.
Used with the GasPak™ EZ Gas Generating Systems. BD™ GasPak™ Supplies are used with these complete, integrated systems for rapid, waterless and catalyst-free gas generation. Can be used to support an anaerobic, microaerophilic or CO2 enriched environment.
Thermo Scientific™ Oxoid™ Resazurin Anaerobic Indicator Available on GSA/VA Contract for Federal Government customers only.
Oxoid Resazurin is an indicator strip for use with anaerobic jar systems.
BD Bio-Bag™ Environmental Chambers Available on GSA/VA Contract for Federal Government customers only.
Disposable systems for cultivation and isolation of specific bacteria and cultures
Thermo Scientific™ Oxoid™ AnaeroGen™ 3.5L Sachet Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Oxoid AnaeroGen 3.5L Sachet is for use with Thermo Scientific™ Oxoid Anaerobic 3.5L jars, Part No. HP0011A, HP0031A.
Thermo Scientific™ Oxoid™ Anaerobic 3.5L Jar Available on GSA/VA Contract for Federal Government customers only.
Oxoid Anaerobic 3.5L System comes with polycarbonate jar and all necessary components.
Thermo Scientific™ Oxoid™ CampyGen™ 2.5L Sachet Available on GSA/VA Contract for Federal Government customers only.
Produce suitable gaseous atmosphere for growth of microaerophilic microorganisms with Oxoid CampyGen 2.5L Sachet.
Thermo Scientific™ AnaeroPouch™-MicroAero Gas Generator Available on GSA/VA Contract for Federal Government customers only.
Mitsubishi™ AnaeroPouch-MicroAero is used for cultivation of microaerophilic microorganisms w/o water.
Thermo Scientific™ Oxoid™ CampyGen™ 3.5L Sachet
Produce suitable gaseous atmosphere for growth of microaerophilic microorganisms with the Oxoid CampyGen 3.5L Sachet.
BD GasPak™ EZ Gas Generating Systems Incubation Containers
Produce atmospheres suitable to support the primary isolation and cultivation of a wide range of bacteria. BD Difco™ GasPak™ EZ Gas Generating Systems Incubation Containers are multi-use systems able to support the primary isolation and cultivation of anaerobic, microaerophilic, or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.
Thermo Scientific™ Oxoid™ CO2 Gen™ Sachet
Generate carbon dioxide atmosphere in AnaeroJar™ 2.5L with Oxoid CO2 Gen Sachet.
Thermo Scientific™ AnaeroPouch™-Bag Available on GSA/VA Contract for Federal Government customers only.
Mitsubishi™ AnaeroPouch-Bag is used to hold AnaeroPouch-Anaero, AnaeroPouch-MicroAero, and AnaeroPouch-CO2 gas generators.
Thermo Scientific™ AnaeroPouch™-CO2 Generator Available on GSA/VA Contract for Federal Government customers only.
Mitsubishi™ AnaeroPouch-CO2 is used for cultivation of CO2 dependent microorganisms
Thermo Scientific™ Double Zip AnaeroPouch™-Bag Available on GSA/VA Contract for Federal Government customers only.
Mitsubishi™ Double Zip AnaeroPouch-Bag is used to hold AnaeroPouch-Anaero, AnaeroPouch-MicroAero, and AnaeroPouch-CO2 gas generators.
BD GasPak™ 100 Systems Available on GSA/VA Contract for Federal Government customers only.
Vented and non-vented models
Thermo Scientific™ AnaeroPouch™-Keep System Available on GSA/VA Contract for Federal Government customers only.
Reduce prepared culture media to less than 0.1% oxygen in 8 hours with an oxygen-absorbing system for use in cultivation of anaerobic bacteria.
Thermo Scientific™ AnaeroPack™ Rectangular Jar Lid Available on GSA/VA Contract for Federal Government customers only.
Mitsubishi™ AnaeroPack Rectangular Jar Lid is a replacement part for anaerobic jar.
BD CO2 Gas Generators Available on GSA/VA Contract for Federal Government customers only.
For use with Bio-Bag™ Type C Environmental Chambers
  spinner