DNA Polymerases
Thermo Scientific™ DreamTaq Green PCR Master Mix (2X) Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific DreamTaq Green PCR Master Mix (2X) is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq Green buffer, MgCl2, and dNTPs.
Thermo Scientific™ Maxima™ H Minus Reverse Transcriptase Available on GSA/VA Contract for Federal Government customers only.
A highly thermostable, high processivity reverse transcriptase lacking RNase H activity.
Thermo Scientific™ DreamTaq Green PCR Master Mix (2X) Available on GSA/VA Contract for Federal Government customers only.
Save time in PCR applications with Master Mix and get higher sensitivity, longer PCR products and higher yields compared to conventional Taq DNA polymerase.
Thermo Scientific™ DreamTaq DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
Get higher sensitivity, longer PCR products and higher yields in all standard PCR applications with this enhanced Taq DNA polymerase.
Invitrogen™ SuperScript™ IV Reverse Transcriptase Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
A proprietary MMLV mutant with superior robustness and reliability in RT reactions.
Thermo Scientific™ phi29 DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
Perform highly efficient isothermal DNA amplification with phi29 DNA Polymerase, a highly processive polymerase with strong strand displacement activity.
Thermo Scientific™ Maxima Reverse Transcriptase Available on GSA/VA Contract for Federal Government customers only.
Increase synthesis rates with Thermo Scientific™ Maxima™ Reverse Transciptase, an advanced enzyme developed through in vitro evolution of M-MuLV RT.
Thermo Scientific™ Taq DNA Polymerase, Recombinant Available on GSA/VA Contract for Federal Government customers only.
Optimize routine PCR with this highly thermostable DNA polymerase.
Promega M-MLV Reverse Transcriptase
RNA-dependent DNA polymerase can be used in primer extension or cDNA synthesis with >5kb mRNA templates
Thermo Scientific™ DreamTaq Green DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
Save time in standard PCR applications with this enhanced Taq DNA polymerase, supplied with colored buffer enabling direct gel loading.
Promega GoTaq™ Flexi DNA Polymerase
Combines robust amplification, streamlined downstream analysis, and the flexibility to optimize Mg2+ concentration
Promega™ GoTaq™ G2 Green Master Mix
Designed for enhanced amplification
Promega AMV Reverse Transcriptase
Catalyzes the polymerization of DNA using template DNA, RNA, or RNA:DNA hybrids
Fisher BioReagents™ Taq DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
Allows up to 100 cycles of amplification without loss of activity
Thermo Scientific™ dsDNase Available on GSA/VA Contract for Federal Government customers only.
dsDNase is a heat inactivatable recombinant shrimp DNase used to remove genomic DNA contamination of RNA samples before reverse transcription.
Invitrogen™ Platinum™ Taq Green Hot Start DNA Polymerase Non-distribution product as customer accommodation.
Invitrogen™ Platinum™ Taq DNA Polymerase is a convenient and reliable “hot start” thermostable DNA polymerase for PCR that provides enhanced specificity over that of Taq DNA Polymerase.
Thermo Scientific™ RevertAid™ Reverse Transcriptase Available on GSA/VA Contract for Federal Government customers only.
Perform efficient synthesis of full-length first strand cDNA up to 13 kb with this genetically modified M-MuLV RT for routine reverse transcription.
Thermo Scientific™ T4 DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
Ensure accuracy in DNA synthesis with template-dependent T4 DNA Polymerase that catalyzes the 5'-3' synthesis from primed single-stranded DNA.
Promega™ GoScript™ Reverse Transcriptase
Delivers robust, reliable cDNA synthesis of a full range of rare and abundant transcripts, even in the presence of inhibitors
Thermo Scientific™ DreamTaq™ DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ DreamTaq DNA Polymerase is an enhanced TaqDNA polymerase optimized for all standard PCR applications.
Thermo Scientific™ RevertAid H Minus Reverse Transcriptase Available on GSA/VA Contract for Federal Government customers only.
A recombinant M-MuLV RT lacking RNase H activity for high yield cDNA synthesis.
Thermo Scientific™ Bsm DNA Polymerase, Large Fragment Available on GSA/VA Contract for Federal Government customers only.
Optimize applications in which a strong strand displacement activity is needed with this DNA polymerase with a high functional similarity to Bst DNA Polymerase.
Promega™ M-MLV Reverse Transcriptase, RNase H Minus, Point Mutant
Reduce costs compared to SuperScript™ II RT while maintaining performance.
Promega™ GoTaq™ G2 Flexi DNA Polymerase
Amplifies a wide range of PCR templates
ADS Biotec™ Optimase™ DNA Polymerase
Ideal for the detection of low levels of mutation in wild type/mutant DNA heterogenous samples and other high-fidelity PCR applications
Promega™ GoTaq™ G2 DNA Polymerase
Amplifies wide range of PCR templates and provides high-performing results
Thermo Scientific™ T7 DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
Catalyze DNA synthesis in the 5'=>3' direction with T7 DNA Polymerase, a template dependent DNA polymerase.
  spinner