Long Range PCR Reagents and Kits
Thermo Scientific™ Extensor Long PCR Enzyme Mix Products
Amplify long PCR products up to 20kb with this synergistically-acting PCR enzyme blend of Taq DNA polymerase and a proprietary proofreading enzyme.
Thermo Scientific™ DyNAzyme EXT DNA Polymerase
Amplify templates up to 40 kb, difficult templates, and GC-rich templates with this optimized mixture of DyNAzyme II DNA Polymerase and a proofreading enzyme.
  spinner