qPCR Assays
Applied Biosystems™ Protein Thermal Shift™ Dye Kit Non-distribution product as customer accommodation.
This solution significantly reduces the time and cost associated with screening samples relative to currently available methods.
Applied Biosystems™ Protein Thermal Shift™ Starter Kit Non-distribution product as customer accommodation.
This solution significantly reduces the time and cost associated with screening samples relative to currently available methods.
Invitrogen™ Uracil DNA Glycosylase Non-distribution product as customer accommodation.
Removes uracil residues from sugar moiety of single- and double-stranded DNA without destroying phosphodiester backbone
Invitrogen™ Platinum™ Quantitative PCR SuperMix-UDG Non-distribution product as customer accommodation.
Is a ready-to-use mixture for high-specificity, real-time quantitative DNA amplification, containing UDG to prevent carryover contamination (1,2).
Invitrogen™ ROX Reference Dye Non-distribution product as customer accommodation.
ROX Reference Dye is designed to normalize the fluorescent reporter signal in real-time quantitative PCR or RT-PCR.
Invitrogen™ Platinum™ Quantitative PCR SuperMix-UDG With ROX Non-distribution product as customer accommodation.
qPCR SuperMixes with ROX for ABI instruments are ready-to-use reaction mixes for high-specificity, real-time amplification of cDNA, genomic DNA or plasmid DNA.
Applied Biosystems™ TaqPath™ Zika Virus Kit (ZIKV) Non-distribution product as customer accommodation.
A simple-to-use, real-time PCR solution for Zika virus RNA detection:
just add RNA. Now with FDA-issued emergency use authorization
Applied Biosystems™ TaqMan™ Arbovirus Triplex Assay (ZIKV/DENV/CHIKV) Non-distribution product as customer accommodation.
A simple solution for dengue, chikungunya, and Zika virus RNA detection: just add RNA.
Applied Biosystems™ TaqMan™ Zika Virus Kit (ZIKV) Non-distribution product as customer accommodation.
TaqMan™ Zika Virus Kit (ZIKV) is designed to detect viral RNA, prepared from urine or serum samples, for Zika virus with Asian lineage.
  spinner