Thermal Cycler Accessories
Applied Biosystems™ TaqMan™ RNase P Instrument Verification Plates Non-distribution product as customer accommodation.
Preloaded with the necessary reagents to detect and quantitate genomic copies of the human RNase P gene.
Applied Biosystems™ StepOnePlus™ Real-Time PCR System Spectral Calibration Kit (96-well) Non-distribution product as customer accommodation.
Easy-to-use kit establishes the pure dye spectra and multicomponent values for FAM™/SYBR™ Green I, VIC™/JOE™, and NED™/TAMRA™/ROX™ dyes on the StepOnePlus™ RT-PCR System.
Applied Biosystems™ Region of Interest (ROI) and Background Plates, 96-well Non-distribution product as customer accommodation.
Used to maintain the ViiA™ 7, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6, QuantStudio™ 7, and QuantStudio™ 12K Flex real-time PCR systems with 96-well (0.2 mL) blocks.
Applied Biosystems™ Region of Interest (ROI) and Background Plates, Fast 96-well Non-distribution product as customer accommodation.
Used to maintain the ViiA™ 7, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6, QuantStudio™ 7, and QuantStudio™ 12K Flex real-time PCR systems with Fast 96-well (0.1mL) blocks.
Applied Biosystems™ Array Card Calibration Install Kit Non-distribution product as customer accommodation.
This kit is used to maintain your ViiA 7 System with TaqMan™ Array Card Block.
Applied Biosystems™ Region of Interest (ROI) and Background Plates, 384-well Non-distribution product as customer accommodation.
Region-of-Interest (ROI) and Background Plates are used to maintain the ViiA™ 7, QuantStudio™ 5, QuantStudio™ 6, QuantStudio™ 7 & QuantStudio™ 12K Flex real-time PCR systems.
Applied Biosystems™ StepOne™ Real-Time PCR System Spectral Calibration Kit (48 well) Non-distribution product as customer accommodation.
Easy-to-use kit establishes the pure dye spectra and multicomponent values for FAM™/SYBR™ Green I, VIC™/JOE™, and ROX™ dyes on the StepOne™ Real-Time PCR System.
Applied Biosystems™ SYBR™ Green Dye Spectral Calibration Plate, 384-well Non-distribution product as customer accommodation.
Used for SYBR™ spectral calibration of your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, or QuantStudio™ 12K Flex system with a 384-well block.
Applied Biosystems™ FAM™/ROX™ and VIC™/ROX™ Dye Normalization Plates, 384-well Non-distribution product as customer accommodation.
Used for normalization calibration of your ViiA™ 7 System with a 384 Well Block.
Applied Biosystems™ FAM™/ROX™ and VIC™/ROX™ Dye Normalization Plates, Fast 96-well Non-distribution product as customer accommodation.
These normalization plates can be used to maintain your ViiA™ 7 System with Fast 96-well Block.
Applied Biosystems™ Normalization Kit, Fast 96-well Non-distribution product as customer accommodation.
Used to maintain your real-time PCR instrument with Fast 96-well block.
Applied Biosystems™ Normalization Plates With FAM™/ROX™ and VIC™/ROX™ dyes, 96-well Non-distribution product as customer accommodation.
These normalization plates can be used to maintain your ViiA™ 7 System with 96-well Block.
Applied Biosystems™ SYBR™ Green Dye Spectral Calibration Plate, 96-well Non-distribution product as customer accommodation.
Used to maintain your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, or QuantStudio™ 12K Flex system with 96-well block.
Applied Biosystems™ FAM™ Dye Spectral Calibration Plate, 384-well Non-distribution product as customer accommodation.
Used for FAM™ spectral calibration of your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, or QuantStudio™ 12K Flex system with a 384-well block.
Applied Biosystems™ ABY™ Dye Spectral Calibration Plate, 96-well Non-distribution product as customer accommodation.
Used for ABY™ spectral calibration of your Applied Biosystems™ 7500 or ViiA™ 7 real-time PCR system with a 96-well block.
Applied Biosystems™ TAMRA™ Dye Spectral Calibration Plate, Fast 96-well Non-distribution product as customer accommodation.
Used to maintain your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, or QuantStudio™ 12K Flex system with Fast 96-well block.
Applied Biosystems™ FAM™ Dye Spectral Calibration Plate, Fast 96-well Non-distribution product as customer accommodation.
Used to maintain your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, or QuantStudio™ 12K Flex system with Fast 96-well block.
Roche Diagnostics LightCycler™ Capillaries Non-distribution product as customer accommodation.
Reaction vessels for LightCycler™ Carousel-Based System Instruments
Applied Biosystems™ SYBR™ Green Dye Spectral Calibration Plate, Fast 96-well Non-distribution product as customer accommodation.
Used to maintain your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex or QuantStudio™ 12K Flex system with Fast 96-well block.
Applied Biosystems™ MUSTANG PURPLE™ Dye Spectral Calibration Plate, 96-well Non-distribution product as customer accommodation.
Used for MUSTANG PURPLE™ spectral calibration of your 7500, ViiA™ 7, QuantStudio™ 6, QuantStudio™ 7, or QuantStudio™ 12K Flex RT-PCR system with a 96-well block.
Applied Biosystems™ FAM™ Dye Spectral Calibration Plate, 96-well Non-distribution product as customer accommodation.
Used to maintain your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, or QuantStudio™ 12K Flex system with 96-well block.
Applied Biosystems™ VIC™ Dye Spectral Calibration Plate, 96-well Non-distribution product as customer accommodation.
Can be used to maintain your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, or QuantStudio™ 12K Flex system with 96-well block.
Applied Biosystems™ ROX™ Dye Spectral Calibration Plate, 96-well Non-distribution product as customer accommodation.
Used to maintain your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, or QuantStudio™ 12K Flex system with 96-well block.
Applied Biosystems™ TAMRA™ Dye Spectral Calibration Plate, 96-well Non-distribution product as customer accommodation.
Used to maintain your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, or QuantStudio™ 12K Flex system with 96-well block.
Applied Biosystems™ NED™ Dye Spectral Calibration Plate, 96-well Non-distribution product as customer accommodation.
Used to maintain your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, or QuantStudio™ 12K Flex system with 96-well block.
Applied Biosystems™ VIC™ Dye Spectral Calibration Plate, 384-well Non-distribution product as customer accommodation.
Used for VIC™ spectral calibration of your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, or QuantStudio™ 12K Flex system with a 384-well block.
Applied Biosystems™ NED™ Dye Spectral Calibration Plate, 384-well Non-distribution product as customer accommodation.
Used for NED™ spectral calibration of your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, or QuantStudio™ 12K Flex system with a 384-well block.
Applied Biosystems™ VIC™ Dye Spectral Calibration Plate, Fast 96-well Non-distribution product as customer accommodation.
Can be used to maintain your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, or QuantStudio™ 12K Flex system with Fast 96-well block.
Applied Biosystems™ ROX™ Dye Spectral Calibration Plate, 384-well Non-distribution product as customer accommodation.
Used for ROX™ spectral calibration of your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, or QuantStudio™ 12K Flex system with a 384-well block.
Applied Biosystems™ ROX™ Dye Spectral Calibration Plate, Fast 96-well Non-distribution product as customer accommodation.
Used to maintain your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, or QuantStudio™ 12K Flex system with Fast 96-well block.
  spinner