Shipping Labels
Fisherbrand™ Colored Labeling Tape, Rainbow Pack Available on GSA/VA Contract for Federal Government customers only. Get IT Item
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
Brady™ FREEZERBONDZ™ Polyester Thermal Transfer Printer Labels Available on GSA/VA Contract for Federal Government customers only. Get IT Item
Made of durable Freezerbondz™ Polyester (B-492). FreezerBondz Thermal Transfer Printer Labels are designed for laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
Brady™ FreezerBondz II Polyester Laboratory Labels B492 Available on GSA/VA Contract for Federal Government customers only.
FreezerBondz II White Polyester Laboratory Labels are durable polyester markers designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
Brady™ IDXPERT™ and LABXPERT™ FreezerBondz II Labels: Rectangular Available on GSA/VA Contract for Federal Government customers only.
Labels withstand environments from LN2 to autoclave, helping preserve research while maintaining sample traceability
Brady™ Polyester Printable Labels
Self-laminating polyester (B-361) Labels for Desktop Laser Printers are supplied with a white printable or write-on zone and a translucent overlaminating area to protect graphics after printing is complete.
Brady™ BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL, clear polyester film tape Available on GSA/VA Contract for Federal Government customers only. Get IT Item
Use this film tape for a variety of labeling applications. BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL are ideal for centrifuge, Eppendorf and PCR tubes; conical and large tubes; cryo tubes and vials.
Fisherbrand™ Micryo™ Strips and Dots for Cryo Storage, Laser Printer Sheets Available on GSA/VA Contract for Federal Government customers only. Get IT Item
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications.
Brady™ B30 Series Heavy-Duty Printer Ribbon Available on GSA/VA Contract for Federal Government customers only.
B30 Series “Drop & Click” print ribbons enable 5-second ribbon supply changes. No calibrating, no threading, no waste, no hassles and the ink always faces the right direction!
Brady™ FreezerBondz Thermal Transfer Printer Rectangular Labels Available on GSA/VA Contract for Federal Government customers only. Get IT Item
Perform well in common laboratory environments such as freezing spray
Fisherbrand™ Colored Labeling Tape, 1 in. Wide Available on GSA/VA Contract for Federal Government customers only. Get IT Item
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
Fisherbrand™ Colored Labeling Tape Available on GSA/VA Contract for Federal Government customers only. Get IT Item
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
Brady™ Thermal Transfer Printable Label Ribbon Available on GSA/VA Contract for Federal Government customers only. Get IT Item
Thermal Transfer Printable Label Ribbon is used with Brady Thermal Transfer Printable Labels.
Brady™ FreezerBondz Thermal Transfer Printer Rectangular Labels PROMO
Durable Freezerbondz™ Polyester (B-492) markers are designed for laboratory identification applications, such as vials, centrifuge, tubes, test tubes, straws and slides.
Brady™ LABXPERT™ Nylon Cloth Labels Available on GSA/VA Contract for Federal Government customers only.
LABXPERT™ Nylon Cloth Labels are nylon cloth (B-499) wrap-around labels that are highly pliable to conform to tightly curved surfaces such as wire and cable.
Brady™ Thermal Transfer Centrifuge and Vial Labels
Thermal Transfer Centrifuge and Vial Labels are made of Self-Laminating Polyester (B-461), a clear film with a matte white printable zone that is ideal for cables, laboratory ID or general labeling where graphics need to be protected.
Fisherbrand™ Self-Adhesive Labels Get IT Item
Use these white labels on a variety of surfaces. Fisherbrand™ Self-Adhesive Labels are removable and pressure sensitive. Ideal for a wide range of labeling applications.
Brady™ LABXPERT™ FreezerBondz Labels Available on GSA/VA Contract for Federal Government customers only.
For Brady TLS 2200 and PC Link Printers
Brady™ LABXPERT™ Polyester Slide Labels Available on GSA/VA Contract for Federal Government customers only.
Use these labels for laboratory and healthcare applications. Brady™ LABXPERT™ Polyester Slide Labels are for use with LABXPERT™ and IDXPERT™ printers.
Fisher Science Education™ Colored Labeling Tape 0.5 in x 14 yds Available on GSA/VA Contract for Federal Government customers only. Get IT Item
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
Fisherbrand™ Cryogenic Labels for Cryogenic Storage Available on GSA/VA Contract for Federal Government customers only. Get IT Item
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications. Comes in an easy to use dispenser box with 20 sheets per pack.
Argos Technologies PolarSafe™ Dots for Cryo Storage
Use in temperatures as low as -196°C (-320°F)
Brady™ LABXPERT™ High-Performance Lab Polyester Labels Available on GSA/VA Contract for Federal Government customers only.
LABXPERT™ High-Performance Lab Polyester Labels are made of self-laminating polyester (B-461), a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
Brady™ FreezerBondz Thermal Transfer Printer Labels: Rectangular with Circles
Perform well in common laboratory environments such as freezing spray
Therapak™ UN3373 Biological Substance Category B Labels Get IT Item
Universally recognized marking for shipping biological substances by air
Brady™ Labels for BMP™21 Mobile Printer, ID PAL™ and LABPAL™ Printers Available on GSA/VA Contract for Federal Government customers only. Get IT Item
For general identification, laboratory identification, panel identification and plate
  spinner