Ultrasonic Homogenizer Accessories
Fisherbrand™ Sound Enclosure for Models 505/705 Sonic Dismembrators
Sound Enclosure for Model 505/705 Sonic Dismembrator. Sonic Dismembrators are extremely loud devices that can cause discomfort to the user and anyone nearby. The Sound Enclosure reduces noise by approximately 20 dBa and is made to work with all accessories. In addition to reducing noise, the Sound Enclosure has an internal support rod and converter mounting system. Any Fisherbrand probe or horn will be held safely and securely inside the unit.
Fisherbrand™ Sound Enclosure for Model 50 and 120 Sonic Dismembrator Available on GSA/VA Contract for Federal Government customers only.
Sound Enclosure for Model 50/120 Sonic Dismembrator. Sonic Dismembrators are extremely loud devices that can cause discomfort to the user and anyone nearby. The Sound Enclosure reduces noise by approximately 20 dBa and is made to work with all accessories. In addition to reducing noise, the Sound Enclosure has an internal support rod and converter mounting system. Any Fisherbrand probe or horn will be held safely and securely inside the unit.
Fisherbrand™ Replaceable Tip Probe for Model 505 and 705 Sonic Dismembrator Available on GSA/VA Contract for Federal Government customers only.
1/2 in. Replaceable Tip Probe processing 20 to 250mL volumes
Fisherbrand™ Probe Microtips for Model 50 and 120 Sonic Dismembrator, Probe Available on GSA/VA Contract for Federal Government customers only.
These Fisher Scientific™ Probes are designed for use with Model 50 and Model 120 Sonic Dismembrators.
Fisherbrand™ Cup Horn for Model 505 and 705 Sonic Dismembrator
Cup Horn Accessory for use with Model 505/705 Sonic Dismembrator. Indirectly process multiple small samples (1ml or less) in sealed tubes. Prevent cross contamination.
Fisherbrand™ 1/4 in. Probe for Model 50 and 120 Sonic Dismembrator Available on GSA/VA Contract for Federal Government customers only.
1/4 in. Probe for processing 10 - 50 mL volumes
Branson Ultrasonics™ Microtips for 200 and 400 Watt Sonifiers
Designed to process samples contained in small diameter vessels at extremely high intensity
Branson Ultrasonics™ Sonifier™ Replacement Flat Tip For Tapped Horn
Used with 1/2 in. (12mm) tapped horn for general processing
Fisherbrand™ Stand and Clamp for Model 50 and 120 Sonic Dismembrator Available on GSA/VA Contract for Federal Government customers only.
Stand and Converter clamp accessory for Model 50/120 Sonic Dismembrator. This accessory will properly support the converter and probe assembly during sonication.
Branson Ultrasonics™ Soundproof Enclosure
For use with Sonifier™ Cell Disruptors
Fisherbrand™ Jack Stand Available on GSA/VA Contract for Federal Government customers only.
Ideal for raising samples up to a stationary probe mounted within the enclosure or on a stand.
Fisherbrand™ Cup Horn for Model 120 Sonic Dismembrator Available on GSA/VA Contract for Federal Government customers only.
1.5 in. Diameter Cup Horn for Model 120 Sonic Dismembrator. Process samples indirectly to prevent cross contamination.
Fisherbrand™ Cup Horn Stand for Model 505 and 705 Sonic Dismembrator
Support the weight of the 5.5 in. Cup horn/Converter assembly properly on a bench top with Cup Horn Stand for Model 505/705 Sonic Dismembrator
Branson Ultrasonics™ Sonifier™ Disruptor Horns
For use with Sonifier S-250 and S-450 Cell Disruptors
Fisherbrand™ Temperature Probe Accessory for FB705 Sonicator
For monitoring temperature of sensitive samples
Branson Ultrasonics™ Microtips for Sonifiers SLPe, SLPt, 150E and 150T
Designed to process samples to the smallest diameter vessels.
Fisherbrand™ Stand for Model 505 and 705 Sonic Dismembrator Available on GSA/VA Contract for Federal Government customers only.
Stand and Converter clamp for Model 505/705 Sonic Dismembrator. This accessory will properly support the converter and probe assembly during sonication.
Branson Ultrasonics™ Sonifier Cell Disruptor Accessory, Support Stand With Rod
For use with Sonifier Cell Disruptor. Branson Ultrasonics Sonifier Cell Disruptor Accessory, Support Stand with rod is used for ease of running application.
Fisherbrand™ Foot Switch for Model 50 and 120 Sonic Dismembrator Available on GSA/VA Contract for Federal Government customers only.
Turn the system on and off remotely using the Foot Switch for Model 120/50 Sonic Dismembrator.
Branson Ultrasonics™ Sonifier Cell Disruptor Accessory, Convertor Clamp
For use with Sonifier Cell Disruptor. Branson Ultrasonics Sonifier Cell Disruptor Accessory, Convertor Clamp is used for stand for use in Sonifier applications.
Branson Ultrasonics™ 200- and 400-Watt Sonifiers: Rosette Cooling Cells
Rosette Cooling Cells for Branson 200- and 400-Watt Sonifiers
Qsonica™ Q700/Q500 Accessories: 8 Tube Holder Rack
An excellent solution for securely holding tubes in place when working with Q500/Q700 Sonicators.
Fisherbrand™ Model 705 Sonic Dismembrator Accessory
4 Tip horn for use with 500 or 700 watt sonicator systems; replacement tip also available.
Fisherbrand™ Replacement Converter for Model 505 and 705 Sonic Dismembrator
Replacement 20kHz Converter for Model 505/705 Sonic Dismembrator
Fisherbrand™ Power Cables for Model 505 and 705 Sonic Dismembrator Available on GSA/VA Contract for Federal Government customers only.
Power Cable with European plug for Model 505/705 Sonic Dismembrator
Fisherbrand™ Power Cable for FB50-110 Sonic Dismembrator
Power Cable for FB50-110 Sonic Dismembrator
  spinner