Ultrasonic Homogenizers
Fisherbrand™ Model 120 Sonic Dismembrator 
Programable system ideal for small volume liquid processing, 220V. Includes 1/8 in. probe for processing 0.5-15ml volumes. There are 3 probe size options available which allow for processing 0.2-50mL volumes.
Fisherbrand™ Model 505 Sonic Dismembrator 
High power, programmable system ideal for nanotechnology, cell disruption and homogenization applications, 220V. Includes standard 1/2 in. Replaceable tip probe (#FB4220) for processing 20-250 mL volumes. There are many probe size options available which allow for processing 0.2-1000 mL volumes.
Fisherbrand™ Model 50 Sonic Dismembrator 
Economical, small volume cell disruptor, 220V. Includes 1/8 in. probe for processing 0.5-15ml volumes. There are 3 probe size options available which allow for processing 0.2-50mL volumes.
Fisherbrand™ Model 705 Sonic Dismembrator 
Ultra-high power with advanced programming for all liquid processing applications, 220V. Includes standard 1/2 in. Replaceable tip probe (#FB4220) for processing 20-250 mL volumes. There are many probe size options available which allow for processing 0.2-1000 mL volumes.
Branson Ultrasonics™ Sonifier™ S-450A Analog Ultrasonic Cell Disruptor/Homogenizer
Versitile laboratory unit suitable for a broad range of liquid processing applications.
Qsonica Sonicator Q500 
Popular applications include nanoparticle dispersion, creating emulsions, cell lysis and homogenization
Qsonica Sonicator Q700 
A state-of-the-art touch screen interface offers intuitive control and provides a user-friendly experience
MP Biomedicals™ SuperFastPrep-2™ 
Enables the processing of samples in the field, during sample collection. MP Biomedicals SuperFastPrep-2™ is an attachment for a rotary tool developed for vigorous cell disruption (bead beating). It is a handheld, cordless instrument that is compatible with most 2mL tubes containing lysis matrix.
Branson Ultrasonics™ Sonifier™ SFX250/SFX550 Cell Disruptors
Advanced controls gives more precise control over all critical elements of experiment
Branson Ultrasonics™ Sonifier™ SFX150 Cell Disruptor
Branson Ultrasonics Sonifier™ SFX150 Cell Disruptor with advanced controls gives more precise control over all critical elements of experiment.
MP Biomedicals™ FastPrep™-96 Instrument 
High-speed, digitally controlled benchtop homogenizer for the lysis of biological samples.
Qsonica Sonicator Q125 
The unit is effective for standard cell disruption, DNA/RNA shearing, homogenization and many other applications
Qsonica Sonicator Q55 
Compact and cost-effective ultrasonic processor that will occupy less bench space
Qsonica Q800R3 Sonicator
DNA Shearing Sonicator
Branson Ultrasonics™ Sonifier S-250A Analog Ultrasonic Cell Disruptor/Homogenizer
Versitile laboratory unit suitable for a broad range of liquid processing applications.
DigiLab HydroShear Plus Hydrodynamic DNA Shearing Instrument
Shears DNA to produce long fragments. Store specific protocols for shearing different size DNA strands.