Tissue Grinders
Kimble™ Kontes™ Pellet Pestle™ Cordless Motor Available on GSA/VA Contract for Federal Government customers only.
Produces more complete disaggregation than manual mixing
Kimble™ Kontes™ Pellet Pestle™ Cordless Motor: Replacement Motor Adapter
Replacement parts for Kontes Pellet Pestle Cordless Motor
Kimble™ Kontes™ Potter-Elvehjem Tissue Grinder
Large volume with a finely machined PTFE pestle
PYREX™ Potter-Elvehjem Tissue Grinder with PTFE Pestle Available on GSA/VA Contract for Federal Government customers only.
Used in preparing small samples for pathological and biochemical studies
Axygen™ Tissue Grinder
Fits 1.5 and 2.0mL centrifuge tubes
Kimble™ Kontes™ Potter-Elvehjem Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
With serrated PTFE pestle and graduated tube
Kimble™ Kontes™ Dounce Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Designed for cellular work in which the nucleus remains intact after homogenization
Kimble™ Kontes™ Closed System Sterile Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Minimizes the risk of personal contact with samples
Wheaton™ Tapered Tissue Grinders With PTFE Pestles Available on GSA/VA Contract for Federal Government customers only.
Tapered tissue grinder with a PTFE pestle for soft tissues
Wheaton™ Dounce Tissue Grinders
Ideal for enzyme studies. Wheaton™ Dounce Tissue Grinders are designed to retain a high percentage of cell nuclei, mitochondria, and proteins in soft tissues or cell suspensions.
PYREX™ PTFE Pestle Tissue Grinder with Steel Shaft Available on GSA/VA Contract for Federal Government customers only.
Recommended for soft tissues found in the liver and brain
Wheaton™ Potter-Elvehjem Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Choose grinders with or without plastic safety coating
Wheaton™ Dounce Dura-Grind™ Tissue Grinder Available on GSA/VA Contract for Federal Government customers only.
An alternative to glass tissue grinders
Fisherbrand™ Pellet Pestles™ Available on GSA/VA Contract for Federal Government customers only.
Resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Kimble™ Kontes™ Pellet Pestle™ Available on GSA/VA Contract for Federal Government customers only.
Use to resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Kimble™ Kontes™ All-Glass Potter-Elvehjem Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Designed for motor drive using 788000 Stirrer Adapter
PYREX™ Glass Pestle Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Consist of glass tube and pestle with clearance of 0.15mm
PYREX™ Glass Pestle Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Designed to handle both initial grinding and final homogenization
Kimble™ BioMasherII™ Closed System Micro Tissue Homogenizer Available on GSA/VA Contract for Federal Government customers only.
Ensure efficient grinding of samples by minimizing potential exposure to hazardous materials
Kimble™ Kontes™ Duall™ Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.
Combined conical and cylindrical surfaces improve grinding efficiency
  spinner