Cell Lifters, Scrapers, and Spreaders
Fisherbrand™ Cell Scrapers
Easily harvest cells from tissue culture flasks
Corning™ Cell Lifter Available on GSA/VA Contract for Federal Government customers only.
Designed with a wide beveled edge for cell culture dishes. Corning™ Cell Lifters are an ideal tool for harvesting stem cells and colonies while maintaining high viability.
Fisherbrand™ Cell Lifters
Disposable lifters quickly remove cell layers
Corning™ Cell Scrapers Available on GSA/VA Contract for Federal Government customers only.
Angled swivel blade minimizes cell damage and ensures even contact with growth area. Corning™ Cell Scrapers are ideal for harvesting cells.
MP Biomedicals™ Roll & Grow™ Plating Beads
Safely and easily spread yeast and bacterial cells
Bel-Art™ SP Scienceware™ Cell Lifters Available on GSA/VA Contract for Federal Government customers only.
Economical and reusable tools to scrape cell lysates or remove and harvest tissue cultures from all types of dishes
Micronova™ Hand-Held Scraper Tools Available on GSA/VA Contract for Federal Government customers only.
Seamless design helps prevent bacterial growth while cleaning in controlled environments
Bel-Art™ SP Scienceware™ Long Rigid Scraper
Flat scraper has a straight, chisel-type, scraping edge which may be resharpened
Corning™ Falcon™ Cell Scrapers Available on GSA/VA Contract for Federal Government customers only.
Designed to provide maximum accessibility to growth surfaces. Corning™ Falcon™ Cell Scrapers provide superior scraping ability with dishes, plates, and flasks.
Thermo Scientific™ Nunc™ Cell Scrapers Available on GSA/VA Contract for Federal Government customers only.
Conduct a variety of applications with sterile, nonpyrogenic Thermo Scientific™ Nunc™ Cell Scrapers, which feature an adjustable blade.
Bel-Art™ SP Scienceware™ Scrapers
Smooth, chemical-resistant scrapers useful for removing residues from filters, cleaning containers, mixing and distributing solids in fluids, spreading pastes over large areas
  spinner